Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 110694104-110694377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRDL1
Entrez Gene ID
91851
Gene Name
chordin like 1
Gene Aliases
CHL, MGC1, MGCN, NRLN1, VOPT, dA141H5.1
UniGene
Hs.496587
Gene Chromosome Location
Chr.X: 110673856-110796058 on Build GRCh38
Transcripts
NM_001143981.1 NM_001143982.1 NM_001143983.2 NM_145234.3 XM_005262221.1 XM_005262222.3 XM_005262223.1 XM_005262224.1 XM_017029959.1

SNP Detail

SNP ID Chromosome Location
rs964970142 Chr.X: 110694180-110694180 on Build GRCh38
rs901343270 Chr.X: 110694133-110694133 on Build GRCh38
rs144828326 Chr.X: 110694129-110694129 on Build GRCh38
rs977798353 Chr.X: 110694210-110694210 on Build GRCh38
rs754740138 Chr.X: 110694291-110694291 on Build GRCh38
rs771646599 Chr.X: 110694185-110694185 on Build GRCh38
rs967107277 Chr.X: 110694230-110694230 on Build GRCh38
rs375845300 Chr.X: 110694255-110694255 on Build GRCh38
rs768510182 Chr.X: 110694148-110694148 on Build GRCh38
rs199971828 Chr.X: 110694134-110694134 on Build GRCh38
rs745404697 Chr.X: 110694152-110694153 on Build GRCh38
rs760305817 Chr.X: 110694205-110694205 on Build GRCh38
rs746499873 Chr.X: 110694318-110694318 on Build GRCh38
rs201430666 Chr.X: 110694312-110694312 on Build GRCh38
rs766035500 Chr.X: 110694231-110694231 on Build GRCh38
rs141240795 Chr.X: 110694243-110694243 on Build GRCh38
rs920527397 Chr.X: 110694288-110694288 on Build GRCh38
rs387906714 Chr.X: 110694289-110694289 on Build GRCh38
rs762366691 Chr.X: 110694238-110694238 on Build GRCh38
rs774385354 Chr.X: 110694150-110694150 on Build GRCh38
rs764215061 Chr.X: 110694256-110694256 on Build GRCh38
rs747670774 Chr.X: 110694171-110694171 on Build GRCh38
rs781026512 Chr.X: 110694190-110694190 on Build GRCh38
rs990584750 Chr.X: 110694279-110694279 on Build GRCh38
rs1011555082 Chr.X: 110694168-110694168 on Build GRCh38
rs765757313 Chr.X: 110694244-110694244 on Build GRCh38
rs753144767 Chr.X: 110694249-110694249 on Build GRCh38
rs769731966 Chr.X: 110694340-110694340 on Build GRCh38
rs370452695 Chr.X: 110694269-110694269 on Build GRCh38
rs948972513 Chr.X: 110694300-110694300 on Build GRCh38
rs12688415 Chr.X: 110694315-110694315 on Build GRCh38
rs749056126 Chr.X: 110694285-110694285 on Build GRCh38
rs11554172 Chr.X: 110694267-110694267 on Build GRCh38
rs777477747 Chr.X: 110694316-110694316 on Build GRCh38
rs768059216 Chr.X: 110694277-110694277 on Build GRCh38
rs139479571 Chr.X: 110694143-110694143 on Build GRCh38

View Full Product Details