Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231594020-231594271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf57
Entrez Gene ID
165100
Gene Name
chromosome 2 open reading frame 57
Gene Aliases
-
UniGene
Hs.98104
Gene Chromosome Location
Chr.2: 231592864-231594283 on Build GRCh38
Transcripts
NM_152614.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3426201 c.1127T>C p.M376T Chr.2: 231594078-231594078 on Build GRCh38
5527732 c.1107G>A p.V369V Chr.2: 231594058-231594058 on Build GRCh38
4092350 c.1076G>A p.R359H Chr.2: 231594027-231594027 on Build GRCh38
4894968 c.1093G>A p.E365K Chr.2: 231594044-231594044 on Build GRCh38
4459530 c.1129C>T p.R377C Chr.2: 231594080-231594080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775995630 Chr.2: 231594191-231594191 on Build GRCh38
rs762488054 Chr.2: 231594069-231594069 on Build GRCh38
rs187659178 Chr.2: 231594122-231594122 on Build GRCh38
rs761392583 Chr.2: 231594143-231594143 on Build GRCh38
rs765653613 Chr.2: 231594063-231594063 on Build GRCh38
rs954229150 Chr.2: 231594205-231594205 on Build GRCh38
rs375754022 Chr.2: 231594061-231594061 on Build GRCh38
rs777214430 Chr.2: 231594057-231594057 on Build GRCh38
rs760084402 Chr.2: 231594108-231594108 on Build GRCh38
rs541531991 Chr.2: 231594142-231594142 on Build GRCh38
rs780691714 Chr.2: 231594086-231594086 on Build GRCh38
rs891392353 Chr.2: 231594161-231594161 on Build GRCh38
rs201411003 Chr.2: 231594080-231594080 on Build GRCh38
rs760869633 Chr.2: 231594055-231594055 on Build GRCh38
rs770467479 Chr.2: 231594113-231594113 on Build GRCh38
rs369242616 Chr.2: 231594100-231594100 on Build GRCh38
rs779791860 Chr.2: 231594095-231594095 on Build GRCh38
rs763437310 Chr.2: 231594129-231594129 on Build GRCh38
rs750309130 Chr.2: 231594090-231594090 on Build GRCh38
rs749975245 Chr.2: 231594147-231594147 on Build GRCh38
rs775980228 Chr.2: 231594126-231594126 on Build GRCh38
rs772378721 Chr.2: 231594038-231594038 on Build GRCh38
rs572080638 Chr.2: 231594165-231594165 on Build GRCh38
rs75570293 Chr.2: 231594107-231594107 on Build GRCh38
rs369891953 Chr.2: 231594040-231594040 on Build GRCh38
rs373418104 Chr.2: 231594149-231594149 on Build GRCh38
rs780024716 Chr.2: 231594155-231594155 on Build GRCh38
rs141442188 Chr.2: 231594068-231594068 on Build GRCh38
rs369940327 Chr.2: 231594093-231594093 on Build GRCh38
rs756717219 Chr.2: 231594081-231594081 on Build GRCh38
rs554143090 Chr.2: 231594078-231594079 on Build GRCh38
rs201877259 Chr.2: 231594056-231594056 on Build GRCh38
rs1008381030 Chr.2: 231594173-231594173 on Build GRCh38
rs544512156 Chr.2: 231594236-231594236 on Build GRCh38
rs200636460 Chr.2: 231594174-231594174 on Build GRCh38
rs1021554468 Chr.2: 231594246-231594246 on Build GRCh38
rs771080469 Chr.2: 231594106-231594106 on Build GRCh38
rs899890447 Chr.2: 231594204-231594204 on Build GRCh38
rs766495965 Chr.2: 231594140-231594140 on Build GRCh38
rs139764624 Chr.2: 231594078-231594078 on Build GRCh38
rs374136231 Chr.2: 231594160-231594160 on Build GRCh38
rs532618040 Chr.2: 231594214-231594214 on Build GRCh38
rs563782746 Chr.2: 231594181-231594181 on Build GRCh38
rs139496513 Chr.2: 231594105-231594105 on Build GRCh38
rs766973394 Chr.2: 231594144-231594144 on Build GRCh38
rs770786040 Chr.2: 231594141-231594142 on Build GRCh38
rs570750981 Chr.2: 231594099-231594099 on Build GRCh38
rs368883740 Chr.2: 231594164-231594164 on Build GRCh38
rs764367622 Chr.2: 231594138-231594138 on Build GRCh38
rs112363884 Chr.2: 231594159-231594159 on Build GRCh38

View Full Product Details