Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151050672-151050925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC3
Entrez Gene ID
9311
Gene Name
acid sensing ion channel subunit 3
Gene Aliases
ACCN3, DRASIC, SLNAC1, TNaC1
UniGene
Hs.647113
Gene Chromosome Location
Chr.7: 151048292-151052756 on Build GRCh38
Transcripts
NM_004769.3 NM_020321.3 NM_020322.3 NR_046401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181574 c.895C>A p.L299I Chr.7: 151050839-151050839 on Build GRCh38
3762493 c.906C>A p.P302P Chr.7: 151050850-151050850 on Build GRCh38
5997313 c.831G>A p.P277P Chr.7: 151050775-151050775 on Build GRCh38
4836135 c.841G>A p.D281N Chr.7: 151050785-151050785 on Build GRCh38
4921395 c.877C>G p.P293A Chr.7: 151050821-151050821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569621626 Chr.7: 151050726-151050726 on Build GRCh38
rs200466021 Chr.7: 151050750-151050750 on Build GRCh38
rs549763020 Chr.7: 151050720-151050720 on Build GRCh38
rs112149514 Chr.7: 151050829-151050829 on Build GRCh38
rs774073299 Chr.7: 151050813-151050813 on Build GRCh38
rs769888171 Chr.7: 151050727-151050727 on Build GRCh38
rs923930575 Chr.7: 151050812-151050812 on Build GRCh38
rs779775702 Chr.7: 151050889-151050889 on Build GRCh38
rs756905170 Chr.7: 151050775-151050775 on Build GRCh38
rs527543284 Chr.7: 151050694-151050694 on Build GRCh38
rs147577517 Chr.7: 151050816-151050816 on Build GRCh38
rs554483671 Chr.7: 151050781-151050781 on Build GRCh38
rs905732965 Chr.7: 151050866-151050866 on Build GRCh38
rs759568152 Chr.7: 151050826-151050826 on Build GRCh38
rs1052941512 Chr.7: 151050843-151050843 on Build GRCh38
rs745324943 Chr.7: 151050785-151050785 on Build GRCh38
rs769451008 Chr.7: 151050787-151050787 on Build GRCh38
rs760573829 Chr.7: 151050882-151050883 on Build GRCh38
rs369864194 Chr.7: 151050890-151050890 on Build GRCh38
rs548452200 Chr.7: 151050730-151050730 on Build GRCh38
rs775509865 Chr.7: 151050799-151050799 on Build GRCh38
rs780659806 Chr.7: 151050873-151050873 on Build GRCh38
rs144330560 Chr.7: 151050739-151050739 on Build GRCh38
rs776943061 Chr.7: 151050823-151050823 on Build GRCh38
rs771099965 Chr.7: 151050818-151050818 on Build GRCh38
rs577646273 Chr.7: 151050803-151050803 on Build GRCh38
rs773266838 Chr.7: 151050846-151050847 on Build GRCh38
rs11977275 Chr.7: 151050850-151050850 on Build GRCh38
rs764493969 Chr.7: 151050840-151050840 on Build GRCh38
rs773415365 Chr.7: 151050741-151050741 on Build GRCh38
rs760874999 Chr.7: 151050744-151050744 on Build GRCh38
rs900351231 Chr.7: 151050830-151050830 on Build GRCh38
rs776059505 Chr.7: 151050722-151050722 on Build GRCh38
rs200386051 Chr.7: 151050847-151050852 on Build GRCh38
rs753138445 Chr.7: 151050834-151050834 on Build GRCh38
rs963857995 Chr.7: 151050742-151050742 on Build GRCh38
rs778006863 Chr.7: 151050770-151050770 on Build GRCh38
rs3217353 Chr.7: 151050862-151050867 on Build GRCh38
rs1020998630 Chr.7: 151050908-151050908 on Build GRCh38
rs768764308 Chr.7: 151050904-151050904 on Build GRCh38
rs763507427 Chr.7: 151050836-151050836 on Build GRCh38
rs535538081 Chr.7: 151050729-151050729 on Build GRCh38
rs755616788 Chr.7: 151050885-151050885 on Build GRCh38
rs758707703 Chr.7: 151050758-151050758 on Build GRCh38
rs114024820 Chr.7: 151050809-151050809 on Build GRCh38
rs140593742 Chr.7: 151050883-151050883 on Build GRCh38
rs201646704 Chr.7: 151050861-151050861 on Build GRCh38
rs564090081 Chr.7: 151050698-151050698 on Build GRCh38
rs1002620719 Chr.7: 151050879-151050879 on Build GRCh38
rs377265988 Chr.7: 151050753-151050753 on Build GRCh38
rs532748577 Chr.7: 151050719-151050719 on Build GRCh38
rs184252418 Chr.7: 151050696-151050696 on Build GRCh38
rs757667487 Chr.7: 151050857-151050857 on Build GRCh38
rs760281117 Chr.7: 151050907-151050907 on Build GRCh38
rs746657424 Chr.7: 151050774-151050774 on Build GRCh38
rs1030266495 Chr.7: 151050878-151050878 on Build GRCh38

View Full Product Details