Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 82928762-82928987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDIAS
Entrez Gene ID
220042
Gene Name
DNA damage induced apoptosis suppressor
Gene Aliases
C11orf82, noxin
UniGene
Hs.165607
Gene Chromosome Location
Chr.11: 82901695-82934659 on Build GRCh38
Transcripts
NM_145018.3 XM_011544835.1 XM_011544836.2 XM_011544837.1 XM_011544838.2 XM_017017352.1 XM_017017353.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5831897 c.153delA p.N52fs*10 Chr.11: 82928816-82928816 on Build GRCh38
4993745 c.184A>G p.K62E Chr.11: 82928847-82928847 on Build GRCh38
1638985 c.158C>T p.A53V Chr.11: 82928821-82928821 on Build GRCh38
69877 c.179C>T p.S60F Chr.11: 82928842-82928842 on Build GRCh38
467478 c.231T>C p.S77S Chr.11: 82928894-82928894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191016474 Chr.11: 82928821-82928821 on Build GRCh38
rs748838201 Chr.11: 82928892-82928892 on Build GRCh38
rs775818272 Chr.11: 82928939-82928939 on Build GRCh38
rs754409597 Chr.11: 82928906-82928906 on Build GRCh38
rs779906159 Chr.11: 82928877-82928877 on Build GRCh38
rs763114662 Chr.11: 82928948-82928948 on Build GRCh38
rs774674376 Chr.11: 82928951-82928951 on Build GRCh38
rs759907660 Chr.11: 82928903-82928904 on Build GRCh38
rs561386530 Chr.11: 82928792-82928792 on Build GRCh38
rs547199855 Chr.11: 82928870-82928870 on Build GRCh38
rs767269393 Chr.11: 82928856-82928856 on Build GRCh38
rs767355216 Chr.11: 82928957-82928957 on Build GRCh38
rs1050642282 Chr.11: 82928942-82928942 on Build GRCh38
rs889336451 Chr.11: 82928954-82928954 on Build GRCh38
rs773811415 Chr.11: 82928796-82928796 on Build GRCh38
rs777132316 Chr.11: 82928931-82928931 on Build GRCh38
rs746436298 Chr.11: 82928935-82928935 on Build GRCh38
rs764058291 Chr.11: 82928949-82928949 on Build GRCh38
rs751681694 Chr.11: 82928828-82928828 on Build GRCh38
rs146662456 Chr.11: 82928801-82928801 on Build GRCh38
rs536127351 Chr.11: 82928938-82928938 on Build GRCh38
rs757123332 Chr.11: 82928842-82928842 on Build GRCh38
rs775700070 Chr.11: 82928824-82928824 on Build GRCh38
rs201402103 Chr.11: 82928910-82928910 on Build GRCh38
rs771377100 Chr.11: 82928804-82928804 on Build GRCh38
rs370203828 Chr.11: 82928826-82928826 on Build GRCh38
rs372898853 Chr.11: 82928916-82928916 on Build GRCh38
rs200694991 Chr.11: 82928955-82928955 on Build GRCh38
rs763056822 Chr.11: 82928831-82928831 on Build GRCh38
rs552584005 Chr.11: 82928815-82928815 on Build GRCh38
rs530822401 Chr.11: 82928813-82928813 on Build GRCh38
rs774686108 Chr.11: 82928868-82928871 on Build GRCh38
rs771462850 Chr.11: 82928926-82928926 on Build GRCh38
rs771253627 Chr.11: 82928832-82928833 on Build GRCh38

View Full Product Details