Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31115793-31116066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDSN
Entrez Gene ID
1041
Gene Name
corneodesmosin
Gene Aliases
HTSS, HTSS1, HYPT2, PSS, PSS1, S
UniGene
Hs.310958
Gene Chromosome Location
Chr.6: 31115088-31120475 on Build GRCh38
Transcripts
NM_001264.4 NM_014068.2

Gene Symbol
PSORS1C1
Entrez Gene ID
170679
Gene Name
psoriasis susceptibility 1 candidate 1
Gene Aliases
C6orf16, SEEK1
UniGene
Hs.310958
Gene Chromosome Location
Chr.6: 31114831-31140092 on Build GRCh38
Transcripts
NM_001264.4 NM_014068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5696254 c.1558C>T p.L520L Chr.6: 31116057-31116057 on Build GRCh38
74109 c.1572G>C p.E524D Chr.6: 31116043-31116043 on Build GRCh38
3624473 c.1568G>A p.G523E Chr.6: 31116047-31116047 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs896643491 Chr.6: 31115986-31115986 on Build GRCh38
rs938854024 Chr.6: 31115885-31115885 on Build GRCh38
rs370908851 Chr.6: 31115916-31115916 on Build GRCh38
rs147539220 Chr.6: 31115994-31115994 on Build GRCh38
rs941335515 Chr.6: 31115890-31115890 on Build GRCh38
rs200992287 Chr.6: 31116022-31116022 on Build GRCh38
rs779015672 Chr.6: 31116042-31116042 on Build GRCh38
rs775263614 Chr.6: 31116030-31116030 on Build GRCh38
rs117066148 Chr.6: 31115915-31115915 on Build GRCh38
rs748282394 Chr.6: 31116002-31116002 on Build GRCh38
rs36195875 Chr.6: 31115932-31115933 on Build GRCh38
rs1042134 Chr.6: 31115887-31115887 on Build GRCh38
rs769638395 Chr.6: 31116028-31116028 on Build GRCh38
rs768599029 Chr.6: 31116040-31116040 on Build GRCh38
rs377342864 Chr.6: 31115978-31115978 on Build GRCh38
rs369310712 Chr.6: 31116024-31116026 on Build GRCh38
rs3095297 Chr.6: 31115952-31115952 on Build GRCh38
rs1057495800 Chr.6: 31115886-31115886 on Build GRCh38
rs56899166 Chr.6: 31116021-31116023 on Build GRCh38
rs778097936 Chr.6: 31116017-31116017 on Build GRCh38
rs994666747 Chr.6: 31115934-31115934 on Build GRCh38
rs3094217 Chr.6: 31115879-31115879 on Build GRCh38
rs144778683 Chr.6: 31115872-31115872 on Build GRCh38
rs758370258 Chr.6: 31116013-31116013 on Build GRCh38
rs750279345 Chr.6: 31116035-31116035 on Build GRCh38
rs3130981 Chr.6: 31116036-31116036 on Build GRCh38
rs140237714 Chr.6: 31115999-31115999 on Build GRCh38

View Full Product Details