Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32305942-32306169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICD1
Entrez Gene ID
636
Gene Name
BICD cargo adaptor 1
Gene Aliases
BICD
UniGene
Hs.505202
Gene Chromosome Location
Chr.12: 32106736-32383637 on Build GRCh38
Transcripts
NM_001003398.1 NM_001714.2 XM_005253465.3 XM_005253466.2 XM_006719134.3 XM_011520812.2 XM_011520813.1 XM_011520814.2 XM_011520815.2 XM_011520816.2 XM_017019797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
938797 c.860G>T p.G287V Chr.12: 32305977-32305977 on Build GRCh38
693330 c.1005+1G>A p.? Chr.12: 32306123-32306123 on Build GRCh38
4877523 c.827G>T p.G276V Chr.12: 32305944-32305944 on Build GRCh38
5726299 c.826G>A p.G276R Chr.12: 32305943-32305943 on Build GRCh38
6072458 c.859G>T p.G287C Chr.12: 32305976-32305976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370085694 Chr.12: 32306124-32306124 on Build GRCh38
rs41521147 Chr.12: 32306136-32306136 on Build GRCh38
rs147476635 Chr.12: 32305966-32305966 on Build GRCh38
rs780557339 Chr.12: 32305970-32305981 on Build GRCh38
rs746632766 Chr.12: 32305992-32305992 on Build GRCh38
rs771769631 Chr.12: 32306046-32306046 on Build GRCh38
rs758094891 Chr.12: 32305978-32305978 on Build GRCh38
rs762037904 Chr.12: 32306044-32306044 on Build GRCh38
rs753665015 Chr.12: 32306052-32306052 on Build GRCh38
rs768819671 Chr.12: 32306026-32306026 on Build GRCh38
rs771218516 Chr.12: 32305994-32305994 on Build GRCh38
rs984671151 Chr.12: 32306127-32306127 on Build GRCh38
rs149973200 Chr.12: 32306110-32306110 on Build GRCh38
rs200645288 Chr.12: 32306054-32306054 on Build GRCh38
rs575147257 Chr.12: 32306133-32306133 on Build GRCh38
rs770467374 Chr.12: 32306141-32306141 on Build GRCh38
rs1023297537 Chr.12: 32306137-32306137 on Build GRCh38
rs36092808 Chr.12: 32306062-32306062 on Build GRCh38
rs752243115 Chr.12: 32306063-32306063 on Build GRCh38
rs747435523 Chr.12: 32306131-32306134 on Build GRCh38
rs555237184 Chr.12: 32306069-32306069 on Build GRCh38
rs756864933 Chr.12: 32306131-32306131 on Build GRCh38
rs201853914 Chr.12: 32305999-32305999 on Build GRCh38
rs774463819 Chr.12: 32306030-32306030 on Build GRCh38
rs199502236 Chr.12: 32306016-32306016 on Build GRCh38
rs777284267 Chr.12: 32306089-32306089 on Build GRCh38
rs746215425 Chr.12: 32306138-32306138 on Build GRCh38
rs780741207 Chr.12: 32306143-32306143 on Build GRCh38
rs777372640 Chr.12: 32305985-32305985 on Build GRCh38
rs759529511 Chr.12: 32306053-32306053 on Build GRCh38
rs375989586 Chr.12: 32306117-32306117 on Build GRCh38
rs770418190 Chr.12: 32306003-32306003 on Build GRCh38
rs374149547 Chr.12: 32305976-32305976 on Build GRCh38

View Full Product Details