Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7607930-7608187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMSAP3
Entrez Gene ID
57662
Gene Name
calmodulin regulated spectrin associated protein family member 3
Gene Aliases
KIAA1543, NEZHA, PPP1R80
UniGene
Hs.17686
Gene Chromosome Location
Chr.19: 7595902-7618310 on Build GRCh38
Transcripts
NM_001080429.2 NM_020902.1 XM_005272491.3 XM_011528154.2 XM_017027036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3783305 c.655C>T p.R219C Chr.19: 7608159-7608159 on Build GRCh38
1397595 c.651G>A p.A217A Chr.19: 7608155-7608155 on Build GRCh38
3783303 c.736C>T p.R246C Chr.19: 7608159-7608159 on Build GRCh38
1397593 c.732G>A p.A244A Chr.19: 7608155-7608155 on Build GRCh38
4557193 c.637G>A p.D213N Chr.19: 7608141-7608141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767034964 Chr.19: 7608085-7608085 on Build GRCh38
rs1055269709 Chr.19: 7607984-7607984 on Build GRCh38
rs939485333 Chr.19: 7607976-7607976 on Build GRCh38
rs773909989 Chr.19: 7608102-7608102 on Build GRCh38
rs757179247 Chr.19: 7608145-7608145 on Build GRCh38
rs763101819 Chr.19: 7608036-7608036 on Build GRCh38
rs571937717 Chr.19: 7608103-7608103 on Build GRCh38
rs911841695 Chr.19: 7608109-7608109 on Build GRCh38
rs199695046 Chr.19: 7608144-7608144 on Build GRCh38
rs371424762 Chr.19: 7608160-7608160 on Build GRCh38
rs755853522 Chr.19: 7608134-7608134 on Build GRCh38
rs559545477 Chr.19: 7608044-7608044 on Build GRCh38
rs962199021 Chr.19: 7607996-7607996 on Build GRCh38
rs771550989 Chr.19: 7608164-7608164 on Build GRCh38
rs368361887 Chr.19: 7608147-7608147 on Build GRCh38
rs201927154 Chr.19: 7608072-7608072 on Build GRCh38
rs944621431 Chr.19: 7608116-7608116 on Build GRCh38
rs199552187 Chr.19: 7608159-7608159 on Build GRCh38
rs752323641 Chr.19: 7608106-7608106 on Build GRCh38
rs763852626 Chr.19: 7608136-7608136 on Build GRCh38
rs372713856 Chr.19: 7608154-7608154 on Build GRCh38
rs767141676 Chr.19: 7608105-7608105 on Build GRCh38
rs920666059 Chr.19: 7608029-7608029 on Build GRCh38
rs770193714 Chr.19: 7608157-7608157 on Build GRCh38
rs973908738 Chr.19: 7608023-7608023 on Build GRCh38
rs199868688 Chr.19: 7608155-7608155 on Build GRCh38
rs1003748216 Chr.19: 7607988-7607988 on Build GRCh38
rs946751409 Chr.19: 7608017-7608017 on Build GRCh38
rs528725744 Chr.19: 7608064-7608064 on Build GRCh38
rs1015505696 Chr.19: 7607993-7607993 on Build GRCh38
rs377116685 Chr.19: 7608076-7608076 on Build GRCh38

View Full Product Details