Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 2753246-2753483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf38
Entrez Gene ID
153571
Gene Name
chromosome 5 open reading frame 38
Gene Aliases
CEI, IRX2NB
UniGene
Hs.148486
Gene Chromosome Location
Chr.5: 2751940-2758576 on Build GRCh38
Transcripts
NM_001294337.1 NM_001306149.1 NM_001306150.1 NM_178569.3 NR_122125.1 NR_122126.1 XM_005248256.3 XM_005248257.3 XM_011513975.2 XM_011513977.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3854650 c.364C>A p.L122M Chr.5: 2753315-2753315 on Build GRCh38
4667820 c.386C>A p.P129H Chr.5: 2753337-2753337 on Build GRCh38
5500748 c.399C>A p.G133G Chr.5: 2753350-2753350 on Build GRCh38
6616268 c.404+6C>T p.? Chr.5: 2753361-2753361 on Build GRCh38
673685 c.392C>A p.P131Q Chr.5: 2753343-2753343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201767186 Chr.5: 2753275-2753275 on Build GRCh38
rs369888211 Chr.5: 2753387-2753387 on Build GRCh38
rs749847357 Chr.5: 2753404-2753404 on Build GRCh38
rs920742819 Chr.5: 2753365-2753365 on Build GRCh38
rs150419625 Chr.5: 2753330-2753330 on Build GRCh38
rs566728393 Chr.5: 2753434-2753434 on Build GRCh38
rs753312806 Chr.5: 2753379-2753379 on Build GRCh38
rs939340688 Chr.5: 2753382-2753382 on Build GRCh38
rs1024903443 Chr.5: 2753433-2753433 on Build GRCh38
rs371931364 Chr.5: 2753362-2753362 on Build GRCh38
rs763177076 Chr.5: 2753354-2753354 on Build GRCh38
rs967606776 Chr.5: 2753437-2753437 on Build GRCh38
rs756880914 Chr.5: 2753307-2753307 on Build GRCh38
rs199779338 Chr.5: 2753332-2753332 on Build GRCh38
rs759861792 Chr.5: 2753369-2753369 on Build GRCh38
rs778413651 Chr.5: 2753383-2753383 on Build GRCh38
rs751381053 Chr.5: 2753290-2753290 on Build GRCh38
rs571510621 Chr.5: 2753355-2753355 on Build GRCh38
rs768956939 Chr.5: 2753328-2753328 on Build GRCh38
rs777080924 Chr.5: 2753301-2753301 on Build GRCh38
rs767160517 Chr.5: 2753373-2753373 on Build GRCh38
rs375334237 Chr.5: 2753376-2753376 on Build GRCh38
rs533415600 Chr.5: 2753402-2753402 on Build GRCh38
rs940090585 Chr.5: 2753343-2753343 on Build GRCh38
rs745463585 Chr.5: 2753385-2753385 on Build GRCh38
rs140455889 Chr.5: 2753299-2753299 on Build GRCh38
rs1055344463 Chr.5: 2753386-2753386 on Build GRCh38
rs999427071 Chr.5: 2753442-2753442 on Build GRCh38
rs748532943 Chr.5: 2753335-2753335 on Build GRCh38
rs946917095 Chr.5: 2753395-2753395 on Build GRCh38
rs745548753 Chr.5: 2753309-2753309 on Build GRCh38
rs752542915 Chr.5: 2753375-2753375 on Build GRCh38
rs752530179 Chr.5: 2753298-2753298 on Build GRCh38
rs988146465 Chr.5: 2753288-2753288 on Build GRCh38
rs778574867 Chr.5: 2753308-2753308 on Build GRCh38
rs781073468 Chr.5: 2753297-2753297 on Build GRCh38
rs754863877 Chr.5: 2753295-2753295 on Build GRCh38
rs1001852663 Chr.5: 2753450-2753450 on Build GRCh38
rs764140504 Chr.5: 2753378-2753378 on Build GRCh38
rs111354291 Chr.5: 2753421-2753421 on Build GRCh38
rs756383394 Chr.5: 2753289-2753289 on Build GRCh38
rs770403378 Chr.5: 2753341-2753341 on Build GRCh38
rs781402733 Chr.5: 2753397-2753397 on Build GRCh38
rs765162378 Chr.5: 2753379-2753379 on Build GRCh38
rs767920768 Chr.5: 2753346-2753346 on Build GRCh38
rs905815776 Chr.5: 2753423-2753423 on Build GRCh38
rs200810855 Chr.5: 2753280-2753280 on Build GRCh38
rs138224052 Chr.5: 2753349-2753349 on Build GRCh38
rs916813657 Chr.5: 2753394-2753394 on Build GRCh38
rs374466417 Chr.5: 2753399-2753399 on Build GRCh38
rs761804356 Chr.5: 2753354-2753355 on Build GRCh38
rs776536184 Chr.5: 2753351-2753352 on Build GRCh38
rs750161987 Chr.5: 2753405-2753406 on Build GRCh38
rs201678854 Chr.5: 2753350-2753350 on Build GRCh38
rs368202268 Chr.5: 2753287-2753287 on Build GRCh38

View Full Product Details