Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36200757-36201012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRPF3
Entrez Gene ID
27154
Gene Name
bromodomain and PHD finger containing 3
Gene Aliases
-
UniGene
Hs.520096
Gene Chromosome Location
Chr.6: 36196773-36232790 on Build GRCh38
Transcripts
NM_015695.2 XM_005249010.2 XM_005249011.3 XM_011514489.1 XM_011514490.1 XM_011514491.1 XM_011514492.1 XM_017010742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6303518 c.668G>T p.C223F Chr.6: 36200990-36200990 on Build GRCh38
421346 c.559C>T p.L187L Chr.6: 36200881-36200881 on Build GRCh38
3762006 c.529A>G p.S177G Chr.6: 36200851-36200851 on Build GRCh38
5931797 c.666A>T p.E222D Chr.6: 36200988-36200988 on Build GRCh38
5753000 c.630T>C p.D210D Chr.6: 36200952-36200952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771796019 Chr.6: 36200949-36200949 on Build GRCh38
rs1002371608 Chr.6: 36200950-36200950 on Build GRCh38
rs760221177 Chr.6: 36200958-36200958 on Build GRCh38
rs376016984 Chr.6: 36200973-36200973 on Build GRCh38
rs765953683 Chr.6: 36200969-36200969 on Build GRCh38
rs761535196 Chr.6: 36200939-36200939 on Build GRCh38
rs1047819954 Chr.6: 36200888-36200888 on Build GRCh38
rs778546779 Chr.6: 36200843-36200843 on Build GRCh38
rs45504893 Chr.6: 36200851-36200851 on Build GRCh38
rs774578899 Chr.6: 36200932-36200932 on Build GRCh38
rs538247175 Chr.6: 36200826-36200826 on Build GRCh38
rs200981567 Chr.6: 36200891-36200891 on Build GRCh38
rs200565609 Chr.6: 36200837-36200837 on Build GRCh38
rs753713981 Chr.6: 36200833-36200833 on Build GRCh38
rs774103028 Chr.6: 36200814-36200814 on Build GRCh38
rs756136144 Chr.6: 36200828-36200828 on Build GRCh38
rs192361205 Chr.6: 36200831-36200831 on Build GRCh38
rs148621811 Chr.6: 36200920-36200920 on Build GRCh38
rs781252237 Chr.6: 36200874-36200874 on Build GRCh38
rs776017623 Chr.6: 36200904-36200904 on Build GRCh38
rs764335120 Chr.6: 36200799-36200799 on Build GRCh38
rs372614303 Chr.6: 36200820-36200820 on Build GRCh38
rs909824197 Chr.6: 36200890-36200890 on Build GRCh38
rs772873756 Chr.6: 36200957-36200957 on Build GRCh38
rs776133125 Chr.6: 36200884-36200884 on Build GRCh38
rs769122940 Chr.6: 36200921-36200921 on Build GRCh38
rs549624999 Chr.6: 36200784-36200784 on Build GRCh38
rs760389567 Chr.6: 36200786-36200786 on Build GRCh38
rs570880014 Chr.6: 36200818-36200818 on Build GRCh38
rs757415182 Chr.6: 36200865-36200865 on Build GRCh38

View Full Product Details