Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95812801-95813026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP57
Entrez Gene ID
9702
Gene Name
centrosomal protein 57
Gene Aliases
MVA2, PIG8, TSP57
UniGene
Hs.101014
Gene Chromosome Location
Chr.11: 95790461-95832693 on Build GRCh38
Transcripts
NM_001243776.1 NM_001243777.1 NM_014679.4 XM_006718945.2 XM_006718946.2 XM_017018592.1 XM_017018593.1 XM_017018594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1248100 c.254T>G p.L85R Chr.11: 95812983-95812983 on Build GRCh38
1200828 c.241C>T p.R81* Chr.11: 95812970-95812970 on Build GRCh38
6552576 c.256A>T p.K86* Chr.11: 95813012-95813012 on Build GRCh38
6552574 c.218G>T p.R73L Chr.11: 95812974-95812974 on Build GRCh38
6627095 c.274G>A p.E92K Chr.11: 95813003-95813003 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368273217 Chr.11: 95812955-95812955 on Build GRCh38
rs902947683 Chr.11: 95812902-95812902 on Build GRCh38
rs200595794 Chr.11: 95812952-95812952 on Build GRCh38
rs779271922 Chr.11: 95812859-95812859 on Build GRCh38
rs764178959 Chr.11: 95812918-95812918 on Build GRCh38
rs555939478 Chr.11: 95812869-95812869 on Build GRCh38
rs750728317 Chr.11: 95812962-95812962 on Build GRCh38
rs772654284 Chr.11: 95812881-95812881 on Build GRCh38
rs912374722 Chr.11: 95812987-95812987 on Build GRCh38
rs202201480 Chr.11: 95812972-95812972 on Build GRCh38
rs201955574 Chr.11: 95812910-95812910 on Build GRCh38
rs752598778 Chr.11: 95812911-95812911 on Build GRCh38
rs387906977 Chr.11: 95812970-95812970 on Build GRCh38
rs371996019 Chr.11: 95812887-95812887 on Build GRCh38
rs537943906 Chr.11: 95812857-95812857 on Build GRCh38
rs770490978 Chr.11: 95812888-95812888 on Build GRCh38
rs372530656 Chr.11: 95812973-95812973 on Build GRCh38
rs758806662 Chr.11: 95812966-95812966 on Build GRCh38
rs747562420 Chr.11: 95812872-95812872 on Build GRCh38
rs759349394 Chr.11: 95812904-95812904 on Build GRCh38
rs769422229 Chr.11: 95812878-95812878 on Build GRCh38
rs140149031 Chr.11: 95812974-95812974 on Build GRCh38
rs780688123 Chr.11: 95812871-95812871 on Build GRCh38
rs746266583 Chr.11: 95812868-95812868 on Build GRCh38
rs753867312 Chr.11: 95812932-95812932 on Build GRCh38
rs760665731 Chr.11: 95812912-95812912 on Build GRCh38
rs764070988 Chr.11: 95812832-95812832 on Build GRCh38
rs953434975 Chr.11: 95812899-95812900 on Build GRCh38
rs930587293 Chr.11: 95812842-95812842 on Build GRCh38
rs761875185 Chr.11: 95812865-95812868 on Build GRCh38
rs748775990 Chr.11: 95812981-95812981 on Build GRCh38
rs376904579 Chr.11: 95812897-95812897 on Build GRCh38

View Full Product Details