Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129001453-129001690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCC1
Entrez Gene ID
79825
Gene Name
EF-hand and coiled-coil domain containing 1
Gene Aliases
C3orf73, CCDC48
UniGene
Hs.134807
Gene Chromosome Location
Chr.3: 129001604-129040742 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513163.2 XM_011513050.2 XM_017006939.1 XM_017006940.1 XM_017006941.1 XM_017006942.1 XM_017006943.1 XM_017006945.1 XM_017006946.1 XM_017006947.1 XM_017006952.1

Gene Symbol
KIAA1257
Entrez Gene ID
57501
Gene Name
KIAA1257
Gene Aliases
-
UniGene
Hs.518247
Gene Chromosome Location
Chr.3: 128910186-129032431 on Build GRCh38
Transcripts
NM_024768.2 XM_005247781.3 XM_011513163.2 XM_011513050.2 XM_017006939.1 XM_017006940.1 XM_017006941.1 XM_017006942.1 XM_017006943.1 XM_017006945.1 XM_017006946.1 XM_017006947.1 XM_017006952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3765053 c.16A>G p.T6A Chr.3: 129001644-129001644 on Build GRCh38
1199984 c.31G>A p.G11S Chr.3: 129001659-129001659 on Build GRCh38
3392025 c.61C>T p.R21W Chr.3: 129001689-129001689 on Build GRCh38
4769395 c.50G>A p.G17D Chr.3: 129001678-129001678 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547077945 Chr.3: 129001514-129001514 on Build GRCh38
rs999675082 Chr.3: 129001525-129001525 on Build GRCh38
rs938612511 Chr.3: 129001476-129001476 on Build GRCh38
rs948126598 Chr.3: 129001503-129001503 on Build GRCh38
rs971841621 Chr.3: 129001635-129001635 on Build GRCh38
rs1028623359 Chr.3: 129001604-129001604 on Build GRCh38
rs934857536 Chr.3: 129001561-129001561 on Build GRCh38
rs570602884 Chr.3: 129001654-129001654 on Build GRCh38
rs72977188 Chr.3: 129001611-129001611 on Build GRCh38
rs961800230 Chr.3: 129001623-129001623 on Build GRCh38
rs370208626 Chr.3: 129001548-129001548 on Build GRCh38
rs559331172 Chr.3: 129001522-129001522 on Build GRCh38
rs535167034 Chr.3: 129001663-129001663 on Build GRCh38
rs921283972 Chr.3: 129001501-129001501 on Build GRCh38
rs773592981 Chr.3: 129001649-129001649 on Build GRCh38
rs147900895 Chr.3: 129001505-129001505 on Build GRCh38
rs903977916 Chr.3: 129001520-129001520 on Build GRCh38
rs902074396 Chr.3: 129001567-129001567 on Build GRCh38
rs947381450 Chr.3: 129001517-129001517 on Build GRCh38
rs999506884 Chr.3: 129001582-129001593 on Build GRCh38
rs898853214 Chr.3: 129001620-129001620 on Build GRCh38
rs997770367 Chr.3: 129001595-129001595 on Build GRCh38
rs1016410806 Chr.3: 129001619-129001619 on Build GRCh38
rs1052632291 Chr.3: 129001528-129001528 on Build GRCh38
rs1031495612 Chr.3: 129001584-129001595 on Build GRCh38
rs569885346 Chr.3: 129001590-129001590 on Build GRCh38
rs1871951 Chr.3: 129001644-129001644 on Build GRCh38
rs530314755 Chr.3: 129001524-129001524 on Build GRCh38
rs751195044 Chr.3: 129001671-129001671 on Build GRCh38
rs1044190686 Chr.3: 129001507-129001507 on Build GRCh38
rs953156652 Chr.3: 129001607-129001607 on Build GRCh38
rs748862254 Chr.3: 129001637-129001637 on Build GRCh38
rs916569327 Chr.3: 129001497-129001497 on Build GRCh38
rs144678686 Chr.3: 129001481-129001481 on Build GRCh38

View Full Product Details