Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8007839-8008100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABLIM2
Entrez Gene ID
84448
Gene Name
actin binding LIM protein family member 2
Gene Aliases
-
UniGene
Hs.233404
Gene Chromosome Location
Chr.4: 7965310-8158832 on Build GRCh38
Transcripts
NM_001130083.1 NM_001130084.1 NM_001130085.1 NM_001130086.1 NM_001130087.1 NM_001130088.1 NM_001286688.1 NM_032432.4 XM_005248014.4 XM_005248017.4 XM_005248018.4 XM_005248019.4 XM_005248020.4 XM_005248021.4 XM_005248022.4 XM_005248023.4 XM_005248024.4 XM_005248027.4 XM_005248028.4 XM_005248029.4 XM_005248030.4 XM_005248031.4 XM_006713923.3 XM_006713924.3 XM_006713925.3 XM_006713927.3 XM_011513586.2 XM_017008706.1 XM_017008707.1 XM_017008708.1 XM_017008709.1 XM_017008710.1 XM_017008711.1 XM_017008712.1 XM_017008713.1 XM_017008714.1 XM_017008715.1 XM_017008716.1 XM_017008717.1 XM_017008718.1 XM_017008719.1 XM_017008720.1 XM_017008721.1 XM_017008722.1 XM_017008723.1 XM_017008724.1 XM_017008725.1 XM_017008726.1 XM_017008727.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1057988 c.1618C>T p.R540* Chr.4: 8008059-8008059 on Build GRCh38
4807744 c.1519C>T p.R507C Chr.4: 8008059-8008059 on Build GRCh38
5401687 c.1593G>A p.R531R Chr.4: 8008084-8008084 on Build GRCh38
5401689 c.1494G>A p.R498R Chr.4: 8008084-8008084 on Build GRCh38
4807743 c.1393C>T p.Q465* Chr.4: 8008059-8008059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979013130 Chr.4: 8007928-8007928 on Build GRCh38
rs111254474 Chr.4: 8008057-8008057 on Build GRCh38
rs559848318 Chr.4: 8007997-8007997 on Build GRCh38
rs761926987 Chr.4: 8008064-8008064 on Build GRCh38
rs1017599696 Chr.4: 8007914-8007914 on Build GRCh38
rs772164676 Chr.4: 8008073-8008073 on Build GRCh38
rs753181469 Chr.4: 8007912-8007912 on Build GRCh38
rs755557050 Chr.4: 8008020-8008020 on Build GRCh38
rs775367988 Chr.4: 8008046-8008046 on Build GRCh38
rs1016417871 Chr.4: 8008001-8008001 on Build GRCh38
rs745698333 Chr.4: 8007994-8007994 on Build GRCh38
rs750640136 Chr.4: 8008009-8008009 on Build GRCh38
rs368849605 Chr.4: 8008048-8008048 on Build GRCh38
rs1008941669 Chr.4: 8007913-8007913 on Build GRCh38
rs367843191 Chr.4: 8007860-8007860 on Build GRCh38
rs774237089 Chr.4: 8008063-8008063 on Build GRCh38
rs781709196 Chr.4: 8007905-8007905 on Build GRCh38
rs17180256 Chr.4: 8008032-8008032 on Build GRCh38
rs769652566 Chr.4: 8007996-8007996 on Build GRCh38
rs376656738 Chr.4: 8008042-8008042 on Build GRCh38
rs746585144 Chr.4: 8008029-8008029 on Build GRCh38
rs965344501 Chr.4: 8007916-8007916 on Build GRCh38
rs80031929 Chr.4: 8008008-8008008 on Build GRCh38
rs889345631 Chr.4: 8007937-8007937 on Build GRCh38
rs749021058 Chr.4: 8008047-8008047 on Build GRCh38
rs762951544 Chr.4: 8007999-8007999 on Build GRCh38
rs769262429 Chr.4: 8008044-8008044 on Build GRCh38
rs754087060 Chr.4: 8008014-8008014 on Build GRCh38
rs780800251 Chr.4: 8008037-8008037 on Build GRCh38
rs768644419 Chr.4: 8008058-8008058 on Build GRCh38
rs1006489660 Chr.4: 8007988-8007988 on Build GRCh38
rs957387630 Chr.4: 8007865-8007865 on Build GRCh38
rs551366205 Chr.4: 8007882-8007882 on Build GRCh38
rs779552345 Chr.4: 8008025-8008025 on Build GRCh38
rs749546974 Chr.4: 8008011-8008011 on Build GRCh38
rs779539842 Chr.4: 8008045-8008046 on Build GRCh38
rs368037222 Chr.4: 8008004-8008004 on Build GRCh38
rs993993527 Chr.4: 8007877-8007877 on Build GRCh38
rs1026368485 Chr.4: 8007881-8007881 on Build GRCh38
rs16841552 Chr.4: 8008010-8008010 on Build GRCh38

View Full Product Details