Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 84631694-84631967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDS1
Entrez Gene ID
1040
Gene Name
CDP-diacylglycerol synthase 1
Gene Aliases
CDS
UniGene
Hs.654899
Gene Chromosome Location
Chr.4: 84582904-84651340 on Build GRCh38
Transcripts
NM_001263.3 XM_005262687.2 XM_017007648.1 XM_017007649.1 XM_017007650.1 XM_017007651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4795126 c.598C>T p.L200L Chr.4: 84631836-84631836 on Build GRCh38
4126410 c.599T>A p.L200Q Chr.4: 84631837-84631837 on Build GRCh38
33542 c.611A>C p.K204T Chr.4: 84631849-84631849 on Build GRCh38
1718380 c.622C>T p.R208C Chr.4: 84631860-84631860 on Build GRCh38
4126411 c.623G>A p.R208H Chr.4: 84631861-84631861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770044262 Chr.4: 84631878-84631878 on Build GRCh38
rs748497937 Chr.4: 84631829-84631829 on Build GRCh38
rs762779246 Chr.4: 84631781-84631781 on Build GRCh38
rs367571119 Chr.4: 84631792-84631792 on Build GRCh38
rs749953686 Chr.4: 84631798-84631798 on Build GRCh38
rs1056384835 Chr.4: 84631807-84631807 on Build GRCh38
rs764336767 Chr.4: 84631781-84631782 on Build GRCh38
rs754421332 Chr.4: 84631798-84631799 on Build GRCh38
rs911543631 Chr.4: 84631859-84631859 on Build GRCh38
rs774163149 Chr.4: 84631899-84631899 on Build GRCh38
rs192407976 Chr.4: 84631932-84631932 on Build GRCh38
rs113753764 Chr.4: 84631774-84631774 on Build GRCh38
rs111327056 Chr.4: 84631797-84631797 on Build GRCh38
rs201288160 Chr.4: 84631790-84631790 on Build GRCh38
rs571604070 Chr.4: 84631725-84631725 on Build GRCh38
rs779827040 Chr.4: 84631824-84631824 on Build GRCh38
rs758455561 Chr.4: 84631930-84631930 on Build GRCh38
rs149864119 Chr.4: 84631929-84631930 on Build GRCh38
rs778776040 Chr.4: 84631842-84631842 on Build GRCh38
rs778162629 Chr.4: 84631844-84631844 on Build GRCh38
rs765440277 Chr.4: 84631775-84631775 on Build GRCh38
rs761912103 Chr.4: 84631905-84631905 on Build GRCh38
rs770946249 Chr.4: 84631770-84631770 on Build GRCh38
rs554765709 Chr.4: 84631928-84631928 on Build GRCh38
rs944347252 Chr.4: 84631860-84631860 on Build GRCh38
rs547619630 Chr.4: 84631771-84631771 on Build GRCh38
rs763972421 Chr.4: 84631782-84631782 on Build GRCh38
rs375312847 Chr.4: 84631784-84631784 on Build GRCh38
rs41282379 Chr.4: 84631729-84631729 on Build GRCh38
rs758747596 Chr.4: 84631833-84631833 on Build GRCh38
rs536408933 Chr.4: 84631897-84631897 on Build GRCh38
rs371649448 Chr.4: 84631810-84631810 on Build GRCh38
rs757786428 Chr.4: 84631800-84631801 on Build GRCh38
rs767554053 Chr.4: 84631910-84631910 on Build GRCh38
rs775741850 Chr.4: 84631884-84631884 on Build GRCh38
rs745934609 Chr.4: 84631871-84631871 on Build GRCh38
rs771001468 Chr.4: 84631861-84631861 on Build GRCh38
rs747491269 Chr.4: 84631855-84631855 on Build GRCh38
rs762681659 Chr.4: 84631891-84631891 on Build GRCh38
rs750006843 Chr.4: 84631913-84631913 on Build GRCh38
rs775314254 Chr.4: 84631778-84631778 on Build GRCh38
rs756735180 Chr.4: 84631791-84631791 on Build GRCh38

View Full Product Details