Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 53716567-53716828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB3
Entrez Gene ID
51130
Gene Name
ankyrin repeat and SOCS box containing 3
Gene Aliases
ASB-3
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53669980-53787009 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001164165.1

Gene Symbol
GPR75-ASB3
Entrez Gene ID
100302652
Gene Name
GPR75-ASB3 readthrough
Gene Aliases
-
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53670293-53860033 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001164165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6091754 c.736C>G p.Q246E Chr.2: 53716726-53716726 on Build GRCh38
6467308 c.662A>C p.Q221P Chr.2: 53716686-53716686 on Build GRCh38
6091755 c.622C>G p.Q208E Chr.2: 53716726-53716726 on Build GRCh38
24385 c.631_648del18 p.D211_L216delDKATPL Chr.2: 53716700-53716717 on Build GRCh38
1021566 c.754C>T p.H252Y Chr.2: 53716594-53716594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761979321 Chr.2: 53716735-53716735 on Build GRCh38
rs769787066 Chr.2: 53716721-53716721 on Build GRCh38
rs781510978 Chr.2: 53716663-53716663 on Build GRCh38
rs367919037 Chr.2: 53716777-53716777 on Build GRCh38
rs764649542 Chr.2: 53716599-53716599 on Build GRCh38
rs749675675 Chr.2: 53716678-53716678 on Build GRCh38
rs750921707 Chr.2: 53716664-53716664 on Build GRCh38
rs143775666 Chr.2: 53716691-53716691 on Build GRCh38
rs140678678 Chr.2: 53716666-53716666 on Build GRCh38
rs902086420 Chr.2: 53716774-53716774 on Build GRCh38
rs141896262 Chr.2: 53716731-53716731 on Build GRCh38
rs776665710 Chr.2: 53716597-53716597 on Build GRCh38
rs767624069 Chr.2: 53716737-53716737 on Build GRCh38
rs1037912150 Chr.2: 53716623-53716623 on Build GRCh38
rs746929491 Chr.2: 53716726-53716726 on Build GRCh38
rs750835406 Chr.2: 53716673-53716676 on Build GRCh38
rs746655115 Chr.2: 53716778-53716778 on Build GRCh38
rs746464337 Chr.2: 53716695-53716695 on Build GRCh38
rs562466698 Chr.2: 53716650-53716650 on Build GRCh38
rs750536999 Chr.2: 53716740-53716740 on Build GRCh38
rs372141675 Chr.2: 53716781-53716781 on Build GRCh38
rs758730125 Chr.2: 53716759-53716759 on Build GRCh38
rs113240917 Chr.2: 53716699-53716699 on Build GRCh38
rs766862100 Chr.2: 53716787-53716788 on Build GRCh38
rs1046770077 Chr.2: 53716729-53716729 on Build GRCh38
rs139081847 Chr.2: 53716743-53716743 on Build GRCh38
rs752543639 Chr.2: 53716612-53716612 on Build GRCh38
rs371904165 Chr.2: 53716679-53716679 on Build GRCh38
rs141422815 Chr.2: 53716645-53716645 on Build GRCh38
rs543212710 Chr.2: 53716621-53716621 on Build GRCh38
rs368231372 Chr.2: 53716795-53716801 on Build GRCh38
rs770446459 Chr.2: 53716696-53716696 on Build GRCh38
rs764992545 Chr.2: 53716601-53716601 on Build GRCh38
rs763154022 Chr.2: 53716616-53716616 on Build GRCh38
rs776051219 Chr.2: 53716704-53716704 on Build GRCh38
rs755429279 Chr.2: 53716750-53716750 on Build GRCh38
rs780720749 Chr.2: 53716748-53716748 on Build GRCh38
rs376556448 Chr.2: 53716725-53716725 on Build GRCh38
rs764300790 Chr.2: 53716632-53716632 on Build GRCh38
rs771796186 Chr.2: 53716687-53716687 on Build GRCh38
rs200706554 Chr.2: 53716657-53716657 on Build GRCh38
rs754209970 Chr.2: 53716749-53716749 on Build GRCh38
rs146708478 Chr.2: 53716794-53716794 on Build GRCh38
rs759501767 Chr.2: 53716715-53716715 on Build GRCh38
rs372488524 Chr.2: 53716754-53716754 on Build GRCh38
rs368230249 Chr.2: 53716674-53716674 on Build GRCh38

View Full Product Details