Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 36240235-36240496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAR2
Entrez Gene ID
25980
Gene Name
AAR2 splicing factor homolog
Gene Aliases
C20orf4, CGI-23
UniGene
Hs.11314
Gene Chromosome Location
Chr.20: 36236136-36256941 on Build GRCh38
Transcripts
NM_001271874.1 NM_015511.4 XM_006723770.3 XM_011528762.2 XM_011528763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6857416 c.499A>G p.M167V Chr.20: 36240367-36240367 on Build GRCh38
4097993 c.610A>T p.T204S Chr.20: 36240478-36240478 on Build GRCh38
1283789 c.392A>T p.K131M Chr.20: 36240260-36240260 on Build GRCh38
2759599 c.618C>T p.I206I Chr.20: 36240486-36240486 on Build GRCh38
478092 c.369G>C p.G123G Chr.20: 36240237-36240237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141691493 Chr.20: 36240420-36240420 on Build GRCh38
rs149423857 Chr.20: 36240474-36240474 on Build GRCh38
rs778338068 Chr.20: 36240380-36240380 on Build GRCh38
rs374608307 Chr.20: 36240422-36240422 on Build GRCh38
rs779633484 Chr.20: 36240387-36240387 on Build GRCh38
rs201609790 Chr.20: 36240346-36240346 on Build GRCh38
rs1046622376 Chr.20: 36240469-36240469 on Build GRCh38
rs749926117 Chr.20: 36240429-36240429 on Build GRCh38
rs896478501 Chr.20: 36240433-36240433 on Build GRCh38
rs781615324 Chr.20: 36240299-36240299 on Build GRCh38
rs76403264 Chr.20: 36240381-36240381 on Build GRCh38
rs774478093 Chr.20: 36240413-36240413 on Build GRCh38
rs750676328 Chr.20: 36240291-36240291 on Build GRCh38
rs751882156 Chr.20: 36240282-36240282 on Build GRCh38
rs372602653 Chr.20: 36240349-36240349 on Build GRCh38
rs781069168 Chr.20: 36240396-36240396 on Build GRCh38
rs34390439 Chr.20: 36240328-36240328 on Build GRCh38
rs201761855 Chr.20: 36240330-36240330 on Build GRCh38
rs768037630 Chr.20: 36240352-36240352 on Build GRCh38
rs377708694 Chr.20: 36240329-36240329 on Build GRCh38
rs368104861 Chr.20: 36240448-36240448 on Build GRCh38
rs768628432 Chr.20: 36240326-36240326 on Build GRCh38
rs201316006 Chr.20: 36240449-36240449 on Build GRCh38
rs373127299 Chr.20: 36240302-36240302 on Build GRCh38
rs1038239797 Chr.20: 36240415-36240415 on Build GRCh38
rs138947526 Chr.20: 36240407-36240407 on Build GRCh38
rs138517690 Chr.20: 36240399-36240399 on Build GRCh38
rs780790498 Chr.20: 36240471-36240471 on Build GRCh38
rs769126744 Chr.20: 36240386-36240386 on Build GRCh38
rs757956329 Chr.20: 36240430-36240430 on Build GRCh38
rs759876242 Chr.20: 36240351-36240351 on Build GRCh38
rs370233623 Chr.20: 36240416-36240416 on Build GRCh38
rs200929659 Chr.20: 36240406-36240406 on Build GRCh38
rs747984876 Chr.20: 36240398-36240398 on Build GRCh38
rs771729009 Chr.20: 36240313-36240313 on Build GRCh38
rs746800707 Chr.20: 36240388-36240388 on Build GRCh38
rs144623771 Chr.20: 36240348-36240348 on Build GRCh38
rs369464017 Chr.20: 36240318-36240318 on Build GRCh38
rs745475861 Chr.20: 36240310-36240310 on Build GRCh38
rs766799042 Chr.20: 36240260-36240260 on Build GRCh38
rs753104874 Chr.20: 36240353-36240353 on Build GRCh38
rs757481964 Chr.20: 36240275-36240275 on Build GRCh38
rs761105630 Chr.20: 36240421-36240421 on Build GRCh38
rs748957834 Chr.20: 36240345-36240345 on Build GRCh38
rs759834621 Chr.20: 36240414-36240414 on Build GRCh38
rs747959052 Chr.20: 36240472-36240472 on Build GRCh38
rs756638120 Chr.20: 36240354-36240354 on Build GRCh38
rs374203470 Chr.20: 36240325-36240325 on Build GRCh38
rs150337322 Chr.20: 36240285-36240285 on Build GRCh38
rs570444876 Chr.20: 36240301-36240301 on Build GRCh38
rs748581464 Chr.20: 36240292-36240305 on Build GRCh38
rs754678537 Chr.20: 36240456-36240456 on Build GRCh38
rs773445065 Chr.20: 36240334-36240334 on Build GRCh38

View Full Product Details