Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55021171-55021418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALAS2
Entrez Gene ID
212
Gene Name
5'-aminolevulinate synthase 2
Gene Aliases
ALAS-E, ALASE, ANH1, ASB, SIDBA1, XLDPP, XLEPP, XLSA
UniGene
Hs.522666
Gene Chromosome Location
Chr.X: 55009055-55031064 on Build GRCh38
Transcripts
NM_000032.4 NM_001037967.3 NM_001037968.3 XM_005261995.3 XM_011530771.1 XM_017029354.1

SNP Detail

SNP ID Chromosome Location
rs763076832 Chr.X: 55021198-55021198 on Build GRCh38
rs977637650 Chr.X: 55021285-55021285 on Build GRCh38
rs763899022 Chr.X: 55021388-55021391 on Build GRCh38
rs143527666 Chr.X: 55021259-55021259 on Build GRCh38
rs374590896 Chr.X: 55021295-55021295 on Build GRCh38
rs751494525 Chr.X: 55021242-55021242 on Build GRCh38
rs777358271 Chr.X: 55021305-55021305 on Build GRCh38
rs137852301 Chr.X: 55021195-55021195 on Build GRCh38
rs991427592 Chr.X: 55021220-55021220 on Build GRCh38
rs768964326 Chr.X: 55021319-55021319 on Build GRCh38
rs773436997 Chr.X: 55021188-55021188 on Build GRCh38
rs1008429241 Chr.X: 55021201-55021201 on Build GRCh38
rs755781232 Chr.X: 55021302-55021302 on Build GRCh38
rs1037997151 Chr.X: 55021381-55021381 on Build GRCh38
rs780471920 Chr.X: 55021309-55021309 on Build GRCh38
rs45479691 Chr.X: 55021308-55021308 on Build GRCh38
rs777562024 Chr.X: 55021231-55021231 on Build GRCh38
rs752426872 Chr.X: 55021273-55021273 on Build GRCh38
rs1057515972 Chr.X: 55021285-55021285 on Build GRCh38
rs137852308 Chr.X: 55021215-55021215 on Build GRCh38
rs145257440 Chr.X: 55021349-55021349 on Build GRCh38
rs753380176 Chr.X: 55021307-55021307 on Build GRCh38
rs774238944 Chr.X: 55021313-55021313 on Build GRCh38
rs749037026 Chr.X: 55021270-55021270 on Build GRCh38

View Full Product Details