Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129834590-129834851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf78
Entrez Gene ID
51759
Gene Name
chromosome 9 open reading frame 78
Gene Aliases
HCA59, HSPC220, bA409K20.3
UniGene
Hs.278429
Gene Chromosome Location
Chr.9: 129827285-129835293 on Build GRCh38
Transcripts
NM_016520.2 XM_011518780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
384157 c.126G>C p.K42N Chr.9: 129834724-129834724 on Build GRCh38
1674889 c.104G>T p.R35I Chr.9: 129834746-129834746 on Build GRCh38
7175719 c.104G>C p.R35T Chr.9: 129834746-129834746 on Build GRCh38
4856926 c.97G>A p.E33K Chr.9: 129834753-129834753 on Build GRCh38
239106 c.93delG p.E32fs*9 Chr.9: 129834757-129834757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773835491 Chr.9: 129834792-129834792 on Build GRCh38
rs769582785 Chr.9: 129834776-129834776 on Build GRCh38
rs772749042 Chr.9: 129834787-129834787 on Build GRCh38
rs3814548 Chr.9: 129834620-129834620 on Build GRCh38
rs41278736 Chr.9: 129834783-129834783 on Build GRCh38
rs762351981 Chr.9: 129834698-129834698 on Build GRCh38
rs762962586 Chr.9: 129834694-129834694 on Build GRCh38
rs746878845 Chr.9: 129834672-129834672 on Build GRCh38
rs370957981 Chr.9: 129834727-129834727 on Build GRCh38
rs529481261 Chr.9: 129834629-129834629 on Build GRCh38
rs762722188 Chr.9: 129834798-129834798 on Build GRCh38
rs775004755 Chr.9: 129834697-129834697 on Build GRCh38
rs751052761 Chr.9: 129834751-129834751 on Build GRCh38
rs931986198 Chr.9: 129834699-129834699 on Build GRCh38
rs375570750 Chr.9: 129834673-129834673 on Build GRCh38
rs560966247 Chr.9: 129834774-129834776 on Build GRCh38
rs1017187361 Chr.9: 129834651-129834651 on Build GRCh38
rs888139402 Chr.9: 129834789-129834789 on Build GRCh38
rs750888060 Chr.9: 129834713-129834713 on Build GRCh38
rs200525467 Chr.9: 129834682-129834682 on Build GRCh38
rs763831212 Chr.9: 129834672-129834674 on Build GRCh38
rs751107953 Chr.9: 129834799-129834799 on Build GRCh38
rs761073432 Chr.9: 129834716-129834716 on Build GRCh38
rs757906505 Chr.9: 129834668-129834668 on Build GRCh38
rs373823052 Chr.9: 129834769-129834769 on Build GRCh38
rs752301503 Chr.9: 129834796-129834796 on Build GRCh38
rs371644803 Chr.9: 129834802-129834802 on Build GRCh38
rs764849524 Chr.9: 129834722-129834722 on Build GRCh38
rs761981667 Chr.9: 129834786-129834786 on Build GRCh38
rs745657015 Chr.9: 129834680-129834680 on Build GRCh38
rs760973134 Chr.9: 129834793-129834793 on Build GRCh38
rs756736019 Chr.9: 129834815-129834815 on Build GRCh38
rs540092085 Chr.9: 129834780-129834780 on Build GRCh38
rs775089842 Chr.9: 129834674-129834674 on Build GRCh38
rs751150302 Chr.9: 129834678-129834680 on Build GRCh38
rs774632595 Chr.9: 129834785-129834785 on Build GRCh38
rs566348414 Chr.9: 129834767-129834767 on Build GRCh38
rs1051174958 Chr.9: 129834825-129834825 on Build GRCh38
rs745696468 Chr.9: 129834772-129834772 on Build GRCh38
rs775756516 Chr.9: 129834684-129834684 on Build GRCh38
rs368896017 Chr.9: 129834640-129834640 on Build GRCh38
rs766696278 Chr.9: 129834794-129834794 on Build GRCh38
rs187882057 Chr.9: 129834819-129834819 on Build GRCh38
rs762624319 Chr.9: 129834741-129834741 on Build GRCh38
rs750287903 Chr.9: 129834809-129834810 on Build GRCh38
rs767584648 Chr.9: 129834773-129834774 on Build GRCh38
rs757248973 Chr.9: 129834752-129834752 on Build GRCh38
rs375148916 Chr.9: 129834816-129834816 on Build GRCh38
rs112144992 Chr.9: 129834743-129834743 on Build GRCh38
rs147629327 Chr.9: 129834656-129834656 on Build GRCh38
rs112226116 Chr.9: 129834776-129834778 on Build GRCh38
rs777715634 Chr.9: 129834670-129834670 on Build GRCh38
rs3814547 Chr.9: 129834742-129834742 on Build GRCh38
rs764195364 Chr.9: 129834696-129834696 on Build GRCh38
rs777202959 Chr.9: 129834747-129834747 on Build GRCh38
rs768125195 Chr.9: 129834708-129834708 on Build GRCh38
rs183155719 Chr.9: 129834817-129834817 on Build GRCh38
rs780170176 Chr.9: 129834779-129834779 on Build GRCh38
rs368566789 Chr.9: 129834690-129834690 on Build GRCh38

View Full Product Details