Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52402173-52402436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAP1
Entrez Gene ID
8314
Gene Name
BRCA1 associated protein 1
Gene Aliases
HUCEP-13, UCHL2, hucep-6
UniGene
Hs.106674
Gene Chromosome Location
Chr.3: 52401004-52410105 on Build GRCh38
Transcripts
NM_004656.3 XM_011534149.2 XM_011534150.2 XM_011534151.2 XM_011534152.1 XM_017007303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6023806 c.2085_2085delC p.N695fs*36 Chr.3: 52402393-52402393 on Build GRCh38
7335243 c.2185C>T p.Q729* Chr.3: 52402293-52402293 on Build GRCh38
6023801 c.2106_2107insA p.G703fs*14 Chr.3: 52402371-52402372 on Build GRCh38
3781660 c.2057-5C>G p.? Chr.3: 52402426-52402426 on Build GRCh38
6939917 c.2167_2174delCCCTACAA p.P723fs*>5 Chr.3: 52402304-52402311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926178223 Chr.3: 52402231-52402231 on Build GRCh38
rs201122466 Chr.3: 52402360-52402360 on Build GRCh38
rs748962519 Chr.3: 52402262-52402262 on Build GRCh38
rs139307137 Chr.3: 52402251-52402251 on Build GRCh38
rs773230722 Chr.3: 52402295-52402295 on Build GRCh38
rs757553269 Chr.3: 52402417-52402417 on Build GRCh38
rs764618711 Chr.3: 52402241-52402241 on Build GRCh38
rs375252451 Chr.3: 52402252-52402252 on Build GRCh38
rs747712653 Chr.3: 52402402-52402402 on Build GRCh38
rs56898787 Chr.3: 52402243-52402243 on Build GRCh38
rs778397771 Chr.3: 52402269-52402269 on Build GRCh38
rs757780962 Chr.3: 52402375-52402375 on Build GRCh38
rs550500585 Chr.3: 52402399-52402399 on Build GRCh38
rs760537008 Chr.3: 52402303-52402303 on Build GRCh38
rs771713346 Chr.3: 52402289-52402289 on Build GRCh38
rs776746400 Chr.3: 52402319-52402319 on Build GRCh38
rs748022925 Chr.3: 52402285-52402285 on Build GRCh38
rs753438711 Chr.3: 52402386-52402386 on Build GRCh38
rs752843061 Chr.3: 52402330-52402330 on Build GRCh38
rs1025761013 Chr.3: 52402215-52402215 on Build GRCh38
rs759611495 Chr.3: 52402320-52402320 on Build GRCh38
rs771007826 Chr.3: 52402314-52402314 on Build GRCh38
rs372987719 Chr.3: 52402383-52402383 on Build GRCh38
rs763400204 Chr.3: 52402357-52402357 on Build GRCh38
rs778647638 Chr.3: 52402390-52402390 on Build GRCh38
rs754513396 Chr.3: 52402387-52402387 on Build GRCh38
rs768488999 Chr.3: 52402264-52402264 on Build GRCh38
rs779640381 Chr.3: 52402256-52402256 on Build GRCh38
rs765539314 Chr.3: 52402328-52402328 on Build GRCh38
rs771901936 Chr.3: 52402412-52402412 on Build GRCh38
rs564062251 Chr.3: 52402228-52402228 on Build GRCh38

View Full Product Details