Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 94628666-94628891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLF1
Entrez Gene ID
84250
Gene Name
SMC5-SMC6 complex localization factor 1
Gene Aliases
ANKRD32, BRCTD1, BRCTx
UniGene
Hs.657315
Gene Chromosome Location
Chr.5: 94613477-94697733 on Build GRCh38
Transcripts
NM_032290.3 XM_017009978.1 XM_017009979.1 XM_017009980.1 XM_017009981.1 XM_017009982.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3947813 c.24T>C p.H8H Chr.5: 94628834-94628834 on Build GRCh38
1071292 c.79C>A p.L27I Chr.5: 94628889-94628889 on Build GRCh38
7355664 c.53A>G p.E18G Chr.5: 94628863-94628863 on Build GRCh38
3141439 c.65C>T p.A22V Chr.5: 94628875-94628875 on Build GRCh38
3141440 c.66G>A p.A22A Chr.5: 94628876-94628876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987249281 Chr.5: 94628703-94628703 on Build GRCh38
rs937312503 Chr.5: 94628755-94628755 on Build GRCh38
rs199976521 Chr.5: 94628821-94628821 on Build GRCh38
rs920157055 Chr.5: 94628806-94628806 on Build GRCh38
rs973388047 Chr.5: 94628796-94628796 on Build GRCh38
rs950197070 Chr.5: 94628705-94628705 on Build GRCh38
rs751690294 Chr.5: 94628774-94628774 on Build GRCh38
rs768328504 Chr.5: 94628745-94628747 on Build GRCh38
rs546232540 Chr.5: 94628723-94628723 on Build GRCh38
rs188220033 Chr.5: 94628860-94628860 on Build GRCh38
rs750772567 Chr.5: 94628843-94628843 on Build GRCh38
rs961579387 Chr.5: 94628795-94628795 on Build GRCh38
rs1015058501 Chr.5: 94628858-94628858 on Build GRCh38
rs905886069 Chr.5: 94628712-94628712 on Build GRCh38
rs757492261 Chr.5: 94628800-94628800 on Build GRCh38
rs559637635 Chr.5: 94628777-94628777 on Build GRCh38
rs1046230604 Chr.5: 94628711-94628711 on Build GRCh38

View Full Product Details