Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117384659-117384920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASZ1
Entrez Gene ID
136991
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Gene Aliases
ALP1, ANKL1, C7orf7, CT1.19, GASZ, Orf3
UniGene
Hs.352412
Gene Chromosome Location
Chr.7: 117363222-117427523 on Build GRCh38
Transcripts
NM_001301821.1 NM_001301822.1 NM_130768.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6441580 c.682C>T p.H228Y Chr.7: 117384731-117384731 on Build GRCh38
4415710 c.647A>C p.K216T Chr.7: 117384766-117384766 on Build GRCh38
6606345 c.564G>A p.W188* Chr.7: 117384849-117384849 on Build GRCh38
3257193 c.671G>T p.R224I Chr.7: 117384742-117384742 on Build GRCh38
6108451 c.613G>A p.G205R Chr.7: 117384800-117384800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772111979 Chr.7: 117384797-117384797 on Build GRCh38
rs766876585 Chr.7: 117384857-117384857 on Build GRCh38
rs748215258 Chr.7: 117384781-117384781 on Build GRCh38
rs768552312 Chr.7: 117384776-117384776 on Build GRCh38
rs765823353 Chr.7: 117384841-117384841 on Build GRCh38
rs779724236 Chr.7: 117384891-117384891 on Build GRCh38
rs190722424 Chr.7: 117384698-117384698 on Build GRCh38
rs760025350 Chr.7: 117384835-117384835 on Build GRCh38
rs766587239 Chr.7: 117384715-117384715 on Build GRCh38
rs924472280 Chr.7: 117384802-117384802 on Build GRCh38
rs751090830 Chr.7: 117384743-117384743 on Build GRCh38
rs766655241 Chr.7: 117384823-117384823 on Build GRCh38
rs761924841 Chr.7: 117384694-117384694 on Build GRCh38
rs377355670 Chr.7: 117384876-117384876 on Build GRCh38
rs749117727 Chr.7: 117384768-117384768 on Build GRCh38
rs573761955 Chr.7: 117384779-117384779 on Build GRCh38
rs148719545 Chr.7: 117384853-117384853 on Build GRCh38
rs776245342 Chr.7: 117384870-117384870 on Build GRCh38
rs755800452 Chr.7: 117384762-117384762 on Build GRCh38
rs756882161 Chr.7: 117384748-117384748 on Build GRCh38
rs770772573 Chr.7: 117384864-117384865 on Build GRCh38
rs749974833 Chr.7: 117384868-117384868 on Build GRCh38
rs755501748 Chr.7: 117384734-117384734 on Build GRCh38
rs777103573 Chr.7: 117384830-117384830 on Build GRCh38
rs773365966 Chr.7: 117384700-117384700 on Build GRCh38
rs143262073 Chr.7: 117384753-117384753 on Build GRCh38
rs543107264 Chr.7: 117384880-117384880 on Build GRCh38
rs373251049 Chr.7: 117384812-117384812 on Build GRCh38
rs534630019 Chr.7: 117384800-117384800 on Build GRCh38
rs1029396 Chr.7: 117384766-117384766 on Build GRCh38
rs780891217 Chr.7: 117384751-117384751 on Build GRCh38
rs761097502 Chr.7: 117384708-117384708 on Build GRCh38
rs753320399 Chr.7: 117384852-117384852 on Build GRCh38
rs765884086 Chr.7: 117384739-117384739 on Build GRCh38

View Full Product Details