Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117534291-117534528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFTR
Entrez Gene ID
1080
Gene Name
cystic fibrosis transmembrane conductance regulator
Gene Aliases
ABC35, ABCC7, CF, CFTR/MRP, MRP7, TNR-CFTR, dJ760C5.1
UniGene
Hs.489786
Gene Chromosome Location
Chr.7: 117478367-117668665 on Build GRCh38
Transcripts
NM_000492.3 XM_011515751.2 XM_011515753.1 XM_011515754.2 XM_017011699.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5871652 c.577G>A p.E193K Chr.7: 117534363-117534363 on Build GRCh38
6254590 c.511G>T p.V171F Chr.7: 117534297-117534297 on Build GRCh38
3632281 c.549C>T p.L183L Chr.7: 117534335-117534335 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1012483278 Chr.7: 117534352-117534352 on Build GRCh38
rs745591601 Chr.7: 117534369-117534369 on Build GRCh38
rs189167747 Chr.7: 117534493-117534493 on Build GRCh38
rs141482808 Chr.7: 117534357-117534357 on Build GRCh38
rs141206145 Chr.7: 117534427-117534427 on Build GRCh38
rs991635265 Chr.7: 117534323-117534323 on Build GRCh38
rs397508750 Chr.7: 117534329-117534332 on Build GRCh38
rs397508751 Chr.7: 117534333-117534333 on Build GRCh38
rs397508754 Chr.7: 117534347-117534347 on Build GRCh38
rs397508756 Chr.7: 117534360-117534360 on Build GRCh38
rs773374503 Chr.7: 117534332-117534332 on Build GRCh38
rs758971927 Chr.7: 117534396-117534396 on Build GRCh38
rs78440224 Chr.7: 117534370-117534370 on Build GRCh38
rs368039301 Chr.7: 117534348-117534348 on Build GRCh38
rs1800080 Chr.7: 117534330-117534330 on Build GRCh38
rs397508755 Chr.7: 117534353-117534353 on Build GRCh38
rs542530565 Chr.7: 117534452-117534452 on Build GRCh38
rs748952496 Chr.7: 117534417-117534417 on Build GRCh38
rs148209839 Chr.7: 117534399-117534399 on Build GRCh38
rs760755991 Chr.7: 117534405-117534405 on Build GRCh38
rs563148024 Chr.7: 117534369-117534369 on Build GRCh38
rs397508752 Chr.7: 117534336-117534336 on Build GRCh38
rs397508757 Chr.7: 117534360-117534362 on Build GRCh38
rs367850319 Chr.7: 117534321-117534321 on Build GRCh38
rs933219867 Chr.7: 117534419-117534419 on Build GRCh38
rs397508759 Chr.7: 117534363-117534363 on Build GRCh38
rs73715571 Chr.7: 117534478-117534478 on Build GRCh38
rs397508753 Chr.7: 117534344-117534344 on Build GRCh38
rs397508761 Chr.7: 117534368-117534368 on Build GRCh38
rs780535019 Chr.7: 117534397-117534397 on Build GRCh38
rs766640075 Chr.7: 117534349-117534349 on Build GRCh38
rs77188391 Chr.7: 117534366-117534366 on Build GRCh38
rs397508758 Chr.7: 117534361-117534361 on Build GRCh38
rs755405930 Chr.7: 117534364-117534364 on Build GRCh38
rs745429476 Chr.7: 117534402-117534402 on Build GRCh38
rs150757811 Chr.7: 117534473-117534473 on Build GRCh38
rs397508760 Chr.7: 117534364-117534370 on Build GRCh38
rs779917088 Chr.7: 117534406-117534406 on Build GRCh38
rs888939277 Chr.7: 117534472-117534472 on Build GRCh38
rs764110352 Chr.7: 117534428-117534428 on Build GRCh38
rs374163420 Chr.7: 117534324-117534324 on Build GRCh38

View Full Product Details