Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46657016-46657289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG13
Entrez Gene ID
9776
Gene Name
autophagy related 13
Gene Aliases
KIAA0652, PARATARG8
UniGene
Hs.127403
Gene Chromosome Location
Chr.11: 46617276-46676019 on Build GRCh38
Transcripts
NM_001142673.2 NM_001205119.1 NM_001205120.1 NM_001205121.1 NM_001205122.1 NM_014741.4 XM_005253262.2 XM_005253263.2 XM_005253265.2 XM_005253266.2 XM_005253268.2 XM_006718394.2 XM_006718395.2 XM_006718396.2 XM_011520493.1 XM_011520496.2 XM_011520499.2 XM_011520500.1 XM_011520501.1 XM_011520502.1 XM_011520503.1 XM_017018599.1 XM_017018600.1 XM_017018601.1 XM_017018602.1 XM_017018603.1 XM_017018604.1 XM_017018605.1 XM_017018606.1 XM_017018607.1 XM_017018608.1 XM_017018609.1 XM_017018610.1 XM_017018611.1 XM_017018612.1 XM_017018613.1 XM_017018614.1 XM_017018615.1 XM_017018616.1 XM_017018617.1 XM_017018618.1 XM_017018619.1 XM_017018620.1 XM_017018621.1 XM_017018622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927248 c.514C>T p.R172C Chr.11: 46657109-46657109 on Build GRCh38
6439896 c.567C>A p.Y189* Chr.11: 46657162-46657162 on Build GRCh38
927247 c.514C>T p.R172C Chr.11: 46657109-46657109 on Build GRCh38
229195 c.536C>T p.P179L Chr.11: 46657131-46657131 on Build GRCh38
3415884 c.515G>A p.R172H Chr.11: 46657110-46657110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764676873 Chr.11: 46657057-46657057 on Build GRCh38
rs781188061 Chr.11: 46657124-46657124 on Build GRCh38
rs369435073 Chr.11: 46657228-46657228 on Build GRCh38
rs755919201 Chr.11: 46657144-46657144 on Build GRCh38
rs201617739 Chr.11: 46657078-46657078 on Build GRCh38
rs914632656 Chr.11: 46657237-46657237 on Build GRCh38
rs748079051 Chr.11: 46657142-46657142 on Build GRCh38
rs953277281 Chr.11: 46657102-46657102 on Build GRCh38
rs751125387 Chr.11: 46657103-46657103 on Build GRCh38
rs189942397 Chr.11: 46657227-46657227 on Build GRCh38
rs770156072 Chr.11: 46657230-46657230 on Build GRCh38
rs780051483 Chr.11: 46657204-46657204 on Build GRCh38
rs1030236252 Chr.11: 46657074-46657074 on Build GRCh38
rs1015271165 Chr.11: 46657045-46657045 on Build GRCh38
rs773712767 Chr.11: 46657237-46657237 on Build GRCh38
rs976024947 Chr.11: 46657059-46657059 on Build GRCh38
rs986360916 Chr.11: 46657104-46657104 on Build GRCh38
rs369264904 Chr.11: 46657061-46657061 on Build GRCh38
rs766619575 Chr.11: 46657241-46657241 on Build GRCh38
rs762205142 Chr.11: 46657072-46657072 on Build GRCh38
rs759630324 Chr.11: 46657109-46657109 on Build GRCh38
rs374721467 Chr.11: 46657046-46657046 on Build GRCh38
rs750047680 Chr.11: 46657082-46657082 on Build GRCh38
rs372693453 Chr.11: 46657183-46657183 on Build GRCh38
rs765916974 Chr.11: 46657091-46657091 on Build GRCh38
rs185314227 Chr.11: 46657198-46657198 on Build GRCh38
rs747080489 Chr.11: 46657216-46657216 on Build GRCh38
rs777570498 Chr.11: 46657153-46657153 on Build GRCh38
rs117856605 Chr.11: 46657041-46657041 on Build GRCh38
rs549075543 Chr.11: 46657120-46657120 on Build GRCh38
rs758150923 Chr.11: 46657088-46657088 on Build GRCh38
rs773618510 Chr.11: 46657240-46657241 on Build GRCh38
rs973820734 Chr.11: 46657072-46657074 on Build GRCh38
rs777070405 Chr.11: 46657070-46657070 on Build GRCh38

View Full Product Details