Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 115165473-115165706 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRNL1
Entrez Gene ID
26033
Gene Name
attractin like 1
Gene Aliases
ALP, bA338L11.1, bA454H24.1
UniGene
Hs.501127
Gene Chromosome Location
Chr.10: 115093365-115948999 on Build GRCh38
Transcripts
NM_001276282.3 NM_207303.4 NR_074088.2 XM_011539587.1 XM_011539588.2 XM_011539589.2 XM_011539590.2 XM_011539591.2 XM_017016035.1 XM_017016036.1 XM_017016037.1 XM_017016038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1317485 c.1005-2A>T p.? Chr.10: 115165556-115165556 on Build GRCh38
6065074 c.1090C>G p.Q364E Chr.10: 115165643-115165643 on Build GRCh38
7357106 c.1091A>T p.Q364L Chr.10: 115165644-115165644 on Build GRCh38
7189623 c.1032T>C p.N344N Chr.10: 115165585-115165585 on Build GRCh38
1703105 c.1043C>T p.P348L Chr.10: 115165596-115165596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782012533 Chr.10: 115165645-115165645 on Build GRCh38
rs925408813 Chr.10: 115165502-115165502 on Build GRCh38
rs782377269 Chr.10: 115165560-115165560 on Build GRCh38
rs192223808 Chr.10: 115165595-115165595 on Build GRCh38
rs782590950 Chr.10: 115165602-115165602 on Build GRCh38
rs782651417 Chr.10: 115165621-115165621 on Build GRCh38
rs781911943 Chr.10: 115165599-115165599 on Build GRCh38
rs949698268 Chr.10: 115165655-115165655 on Build GRCh38
rs782317478 Chr.10: 115165664-115165664 on Build GRCh38
rs782763202 Chr.10: 115165587-115165587 on Build GRCh38
rs370264589 Chr.10: 115165586-115165586 on Build GRCh38
rs782224122 Chr.10: 115165604-115165604 on Build GRCh38
rs782074862 Chr.10: 115165556-115165556 on Build GRCh38
rs781824869 Chr.10: 115165505-115165505 on Build GRCh38
rs782381820 Chr.10: 115165512-115165512 on Build GRCh38
rs781975657 Chr.10: 115165671-115165671 on Build GRCh38
rs200620278 Chr.10: 115165618-115165618 on Build GRCh38
rs781805610 Chr.10: 115165585-115165585 on Build GRCh38
rs782432237 Chr.10: 115165643-115165643 on Build GRCh38
rs782306423 Chr.10: 115165529-115165529 on Build GRCh38
rs782265505 Chr.10: 115165510-115165510 on Build GRCh38
rs782197296 Chr.10: 115165656-115165656 on Build GRCh38
rs782689698 Chr.10: 115165515-115165515 on Build GRCh38
rs782542678 Chr.10: 115165601-115165601 on Build GRCh38
rs782530121 Chr.10: 115165592-115165592 on Build GRCh38
rs147572693 Chr.10: 115165570-115165570 on Build GRCh38
rs377055162 Chr.10: 115165541-115165541 on Build GRCh38
rs782793018 Chr.10: 115165584-115165584 on Build GRCh38
rs782240382 Chr.10: 115165627-115165627 on Build GRCh38

View Full Product Details