Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174808966-174809221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHN1
Entrez Gene ID
1123
Gene Name
chimerin 1
Gene Aliases
ARHGAP2, CHN, DURS2, NC, RHOGAP2
UniGene
Hs.380138
Gene Chromosome Location
Chr.2: 174799314-175005379 on Build GRCh38
Transcripts
NM_001025201.3 NM_001206602.1 NM_001822.5 NR_038133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5734365 c.1006A>G p.I336V Chr.2: 174809001-174809001 on Build GRCh38
6543281 c.1025C>A p.A342E Chr.2: 174808982-174808982 on Build GRCh38
5734364 c.631A>G p.I211V Chr.2: 174809001-174809001 on Build GRCh38
3372494 c.632T>C p.I211T Chr.2: 174809000-174809000 on Build GRCh38
6543280 c.650C>A p.A217E Chr.2: 174808982-174808982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549941864 Chr.2: 174809056-174809056 on Build GRCh38
rs756117866 Chr.2: 174809008-174809008 on Build GRCh38
rs1010121590 Chr.2: 174809185-174809185 on Build GRCh38
rs543766746 Chr.2: 174809179-174809179 on Build GRCh38
rs770666588 Chr.2: 174809026-174809026 on Build GRCh38
rs768856619 Chr.2: 174809042-174809042 on Build GRCh38
rs777550632 Chr.2: 174809009-174809009 on Build GRCh38
rs1047986721 Chr.2: 174809097-174809097 on Build GRCh38
rs371237307 Chr.2: 174809072-174809072 on Build GRCh38
rs778704929 Chr.2: 174809029-174809029 on Build GRCh38
rs535146606 Chr.2: 174809087-174809087 on Build GRCh38
rs763345540 Chr.2: 174809161-174809161 on Build GRCh38
rs576403186 Chr.2: 174809163-174809163 on Build GRCh38
rs531920355 Chr.2: 174808997-174808997 on Build GRCh38
rs943434139 Chr.2: 174809055-174809055 on Build GRCh38
rs1035489342 Chr.2: 174809175-174809175 on Build GRCh38
rs747250069 Chr.2: 174809039-174809039 on Build GRCh38
rs371299944 Chr.2: 174809000-174809000 on Build GRCh38
rs376051698 Chr.2: 174809005-174809005 on Build GRCh38
rs372611145 Chr.2: 174809154-174809154 on Build GRCh38
rs749202508 Chr.2: 174809020-174809020 on Build GRCh38
rs57776382 Chr.2: 174809142-174809142 on Build GRCh38
rs555878429 Chr.2: 174809158-174809158 on Build GRCh38
rs776728291 Chr.2: 174809051-174809051 on Build GRCh38
rs7558477 Chr.2: 174809092-174809092 on Build GRCh38
rs565624744 Chr.2: 174809125-174809125 on Build GRCh38
rs377380143 Chr.2: 174809160-174809160 on Build GRCh38

View Full Product Details