Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96056861-96057094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNJ
Entrez Gene ID
54619
Gene Name
cyclin J
Gene Aliases
bA690P14.1
UniGene
Hs.596479
Gene Chromosome Location
Chr.10: 96042848-96060870 on Build GRCh38
Transcripts
NM_001134375.1 NM_001134376.1 NM_019084.4 XM_005269921.3 XM_011539885.2 XM_017016354.1 XM_017016355.1 XM_017016356.1 NR_038444.1 NR_134320.1 NR_134321.1 NR_134322.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001134375.1 NM_001134376.1 NM_019084.4 XM_005269921.3 XM_011539885.2 XM_017016354.1 XM_017016355.1 XM_017016356.1 NR_038444.1 NR_134320.1 NR_134321.1 NR_134322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4499119 c.534C>T p.L178L Chr.10: 96056954-96056954 on Build GRCh38
5628570 c.441C>T p.A147A Chr.10: 96056861-96056861 on Build GRCh38
231980 c.546A>T p.K182N Chr.10: 96056966-96056966 on Build GRCh38
5837552 c.519_520delGG p.E174fs*3 Chr.10: 96056939-96056940 on Build GRCh38
4499118 c.534C>T p.L178L Chr.10: 96056954-96056954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778821095 Chr.10: 96057047-96057047 on Build GRCh38
rs998607793 Chr.10: 96057004-96057004 on Build GRCh38
rs759076106 Chr.10: 96057022-96057023 on Build GRCh38
rs187113717 Chr.10: 96056918-96056918 on Build GRCh38
rs766794532 Chr.10: 96056984-96056984 on Build GRCh38
rs778391284 Chr.10: 96056901-96056901 on Build GRCh38
rs750048945 Chr.10: 96057049-96057049 on Build GRCh38
rs550221549 Chr.10: 96056928-96056928 on Build GRCh38
rs748195698 Chr.10: 96056967-96056967 on Build GRCh38
rs745546271 Chr.10: 96056909-96056909 on Build GRCh38
rs773779475 Chr.10: 96056965-96056965 on Build GRCh38
rs778099284 Chr.10: 96057059-96057059 on Build GRCh38
rs767673024 Chr.10: 96057033-96057033 on Build GRCh38
rs746455382 Chr.10: 96057056-96057056 on Build GRCh38
rs760851164 Chr.10: 96057036-96057036 on Build GRCh38
rs756954237 Chr.10: 96057046-96057046 on Build GRCh38
rs369388644 Chr.10: 96057035-96057035 on Build GRCh38
rs377087423 Chr.10: 96057045-96057045 on Build GRCh38
rs774385881 Chr.10: 96057016-96057016 on Build GRCh38
rs771553105 Chr.10: 96056914-96056914 on Build GRCh38
rs759884304 Chr.10: 96057023-96057023 on Build GRCh38
rs779529596 Chr.10: 96057055-96057055 on Build GRCh38
rs758117662 Chr.10: 96057051-96057051 on Build GRCh38
rs779153538 Chr.10: 96056893-96056893 on Build GRCh38
rs569953701 Chr.10: 96056966-96056966 on Build GRCh38
rs1011479199 Chr.10: 96057025-96057025 on Build GRCh38
rs748980326 Chr.10: 96056900-96056900 on Build GRCh38
rs763870681 Chr.10: 96057040-96057040 on Build GRCh38
rs770341054 Chr.10: 96056937-96056937 on Build GRCh38

View Full Product Details