Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46601396-46601631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM3
Entrez Gene ID
808
Gene Name
calmodulin 3
Gene Aliases
HEL-S-72, PHKD, PHKD3
UniGene
Hs.515487
Gene Chromosome Location
Chr.19: 46601255-46610782 on Build GRCh38
Transcripts
NM_005184.2

SNP Detail

SNP ID Chromosome Location
rs940660942 Chr.19: 46601518-46601518 on Build GRCh38
rs200606129 Chr.19: 46601460-46601460 on Build GRCh38
rs946956401 Chr.19: 46601539-46601539 on Build GRCh38
rs999014861 Chr.19: 46601425-46601425 on Build GRCh38
rs752910641 Chr.19: 46601474-46601474 on Build GRCh38
rs991337518 Chr.19: 46601453-46601454 on Build GRCh38
rs546364458 Chr.19: 46601504-46601504 on Build GRCh38
rs184847124 Chr.19: 46601522-46601522 on Build GRCh38
rs759824845 Chr.19: 46601450-46601450 on Build GRCh38
rs982176814 Chr.19: 46601454-46601454 on Build GRCh38
rs761428143 Chr.19: 46601449-46601450 on Build GRCh38
rs935366412 Chr.19: 46601512-46601512 on Build GRCh38
rs889267286 Chr.19: 46601550-46601550 on Build GRCh38
rs969897839 Chr.19: 46601432-46601432 on Build GRCh38
rs201296349 Chr.19: 46601421-46601421 on Build GRCh38
rs930846225 Chr.19: 46601596-46601596 on Build GRCh38
rs528529256 Chr.19: 46601529-46601529 on Build GRCh38
rs1022863262 Chr.19: 46601458-46601463 on Build GRCh38
rs879273258 Chr.19: 46601562-46601562 on Build GRCh38
rs774864385 Chr.19: 46601431-46601431 on Build GRCh38
rs1051045394 Chr.19: 46601547-46601547 on Build GRCh38
rs543218237 Chr.19: 46601468-46601468 on Build GRCh38
rs767873951 Chr.19: 46601455-46601455 on Build GRCh38
rs1040159294 Chr.19: 46601600-46601600 on Build GRCh38
rs1018636146 Chr.19: 46601417-46601417 on Build GRCh38
rs548578749 Chr.19: 46601591-46601591 on Build GRCh38

View Full Product Details