Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68865425-68865692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP10
Entrez Gene ID
27302
Gene Name
bone morphogenetic protein 10
Gene Aliases
-
UniGene
Hs.158317
Gene Chromosome Location
Chr.2: 68865481-68871517 on Build GRCh38
Transcripts
NM_014482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4095255 c.1262G>A p.C421Y Chr.2: 68865644-68865644 on Build GRCh38
3055860 c.1251C>T p.A417A Chr.2: 68865655-68865655 on Build GRCh38
6611898 c.1249G>A p.A417T Chr.2: 68865657-68865657 on Build GRCh38
5665949 c.1220T>A p.V407D Chr.2: 68865686-68865686 on Build GRCh38
1409094 c.1252G>A p.V418I Chr.2: 68865654-68865654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529483835 Chr.2: 68865594-68865594 on Build GRCh38
rs761371050 Chr.2: 68865624-68865626 on Build GRCh38
rs768760716 Chr.2: 68865580-68865580 on Build GRCh38
rs906171019 Chr.2: 68865511-68865511 on Build GRCh38
rs759194261 Chr.2: 68865598-68865598 on Build GRCh38
rs112705024 Chr.2: 68865531-68865531 on Build GRCh38
rs1053577869 Chr.2: 68865458-68865458 on Build GRCh38
rs987061866 Chr.2: 68865500-68865500 on Build GRCh38
rs569922324 Chr.2: 68865561-68865561 on Build GRCh38
rs750987492 Chr.2: 68865646-68865646 on Build GRCh38
rs774390280 Chr.2: 68865583-68865583 on Build GRCh38
rs993909043 Chr.2: 68865515-68865515 on Build GRCh38
rs10173546 Chr.2: 68865489-68865489 on Build GRCh38
rs765110764 Chr.2: 68865599-68865599 on Build GRCh38
rs756533986 Chr.2: 68865667-68865667 on Build GRCh38
rs147637852 Chr.2: 68865468-68865468 on Build GRCh38
rs1005679150 Chr.2: 68865603-68865603 on Build GRCh38
rs775636669 Chr.2: 68865610-68865610 on Build GRCh38
rs763748578 Chr.2: 68865629-68865629 on Build GRCh38
rs767015446 Chr.2: 68865654-68865654 on Build GRCh38
rs371379001 Chr.2: 68865635-68865635 on Build GRCh38
rs367847684 Chr.2: 68865623-68865623 on Build GRCh38
rs533477179 Chr.2: 68865508-68865508 on Build GRCh38
rs973838214 Chr.2: 68865556-68865568 on Build GRCh38
rs753505670 Chr.2: 68865557-68865557 on Build GRCh38
rs748423235 Chr.2: 68865592-68865592 on Build GRCh38
rs762235640 Chr.2: 68865487-68865487 on Build GRCh38
rs750817496 Chr.2: 68865655-68865655 on Build GRCh38
rs2231348 Chr.2: 68865649-68865649 on Build GRCh38

View Full Product Details