Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27081003-27081242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf71
Entrez Gene ID
728858
Gene Name
chromosome 12 open reading frame 71
Gene Aliases
-
UniGene
Hs.334003
Gene Chromosome Location
Chr.12: 27081057-27084548 on Build GRCh38
Transcripts
NM_001080406.1 XM_011520847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
430961 c.756C>G p.H252Q Chr.12: 27081228-27081228 on Build GRCh38
3935898 c.772C>G p.Q258E Chr.12: 27081212-27081212 on Build GRCh38
3954595 c.802C>T p.P268S Chr.12: 27081182-27081182 on Build GRCh38
4666805 c.767G>T p.R256I Chr.12: 27081217-27081217 on Build GRCh38
3935899 c.765G>T p.K255N Chr.12: 27081219-27081219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773280490 Chr.12: 27081133-27081133 on Build GRCh38
rs771077595 Chr.12: 27081140-27081140 on Build GRCh38
rs776955026 Chr.12: 27081141-27081141 on Build GRCh38
rs545735885 Chr.12: 27081105-27081105 on Build GRCh38
rs954316257 Chr.12: 27081054-27081054 on Build GRCh38
rs761846945 Chr.12: 27081157-27081157 on Build GRCh38
rs1028571639 Chr.12: 27081042-27081042 on Build GRCh38
rs563920102 Chr.12: 27081124-27081124 on Build GRCh38
rs575177802 Chr.12: 27081092-27081092 on Build GRCh38
rs528311503 Chr.12: 27081125-27081125 on Build GRCh38
rs749924856 Chr.12: 27081192-27081192 on Build GRCh38
rs1012183234 Chr.12: 27081038-27081038 on Build GRCh38
rs377613414 Chr.12: 27081093-27081093 on Build GRCh38
rs767079869 Chr.12: 27081158-27081158 on Build GRCh38
rs768633578 Chr.12: 27081149-27081149 on Build GRCh38
rs528731546 Chr.12: 27081156-27081156 on Build GRCh38
rs762636871 Chr.12: 27081148-27081148 on Build GRCh38
rs779393981 Chr.12: 27081064-27081064 on Build GRCh38
rs1019633370 Chr.12: 27081091-27081091 on Build GRCh38
rs1008192350 Chr.12: 27081063-27081063 on Build GRCh38
rs760337380 Chr.12: 27081209-27081209 on Build GRCh38
rs747852365 Chr.12: 27081139-27081139 on Build GRCh38
rs563200461 Chr.12: 27081062-27081062 on Build GRCh38

View Full Product Details