Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 81063873-81064100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf46
Entrez Gene ID
123775
Gene Name
chromosome 16 open reading frame 46
Gene Aliases
-
UniGene
Hs.658684
Gene Chromosome Location
Chr.16: 81053497-81077267 on Build GRCh38
Transcripts
NM_001100873.1 NM_152337.2 XM_005255792.2 XM_011522848.2 XM_011522849.2 XM_017022916.1 XM_017022917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1709456 c.79C>T p.P27S Chr.16: 81063877-81063877 on Build GRCh38
4623425 c.61T>A p.F21I Chr.16: 81063895-81063895 on Build GRCh38
1709455 c.79C>T p.P27S Chr.16: 81063877-81063877 on Build GRCh38
1679296 c.70G>T p.E24* Chr.16: 81063886-81063886 on Build GRCh38
4063070 c.37A>C p.N13H Chr.16: 81063919-81063919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023630017 Chr.16: 81064034-81064034 on Build GRCh38
rs376166544 Chr.16: 81063961-81063961 on Build GRCh38
rs749934311 Chr.16: 81063986-81063988 on Build GRCh38
rs891519980 Chr.16: 81064017-81064017 on Build GRCh38
rs994153288 Chr.16: 81064055-81064055 on Build GRCh38
rs531893150 Chr.16: 81064030-81064030 on Build GRCh38
rs753311079 Chr.16: 81064001-81064001 on Build GRCh38
rs928313292 Chr.16: 81063898-81063898 on Build GRCh38
rs762306656 Chr.16: 81063969-81063969 on Build GRCh38
rs952902056 Chr.16: 81064061-81064061 on Build GRCh38
rs187543041 Chr.16: 81064042-81064042 on Build GRCh38
rs369420529 Chr.16: 81063920-81063920 on Build GRCh38
rs201201027 Chr.16: 81063949-81063949 on Build GRCh38
rs774878821 Chr.16: 81063966-81063966 on Build GRCh38
rs769062105 Chr.16: 81063965-81063965 on Build GRCh38
rs769930638 Chr.16: 81063931-81063931 on Build GRCh38
rs71400138 Chr.16: 81064040-81064040 on Build GRCh38
rs765989930 Chr.16: 81063998-81063998 on Build GRCh38
rs930088498 Chr.16: 81064016-81064016 on Build GRCh38
rs375358482 Chr.16: 81064005-81064005 on Build GRCh38
rs746079072 Chr.16: 81063921-81063921 on Build GRCh38
rs780005894 Chr.16: 81063941-81063941 on Build GRCh38
rs772540650 Chr.16: 81063989-81063989 on Build GRCh38
rs778454593 Chr.16: 81063977-81063977 on Build GRCh38
rs202168128 Chr.16: 81064003-81064003 on Build GRCh38
rs193278264 Chr.16: 81064056-81064056 on Build GRCh38
rs956872268 Chr.16: 81064053-81064053 on Build GRCh38
rs538738998 Chr.16: 81064043-81064043 on Build GRCh38
rs199990243 Chr.16: 81063993-81063993 on Build GRCh38
rs374995305 Chr.16: 81063908-81063908 on Build GRCh38
rs890080739 Chr.16: 81064033-81064033 on Build GRCh38
rs564667857 Chr.16: 81063995-81063995 on Build GRCh38
rs1011775354 Chr.16: 81064019-81064019 on Build GRCh38
rs1043607170 Chr.16: 81064026-81064026 on Build GRCh38
rs758160285 Chr.16: 81063909-81063909 on Build GRCh38

View Full Product Details