Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15022822-15023077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC105
Entrez Gene ID
126402
Gene Name
coiled-coil domain containing 105
Gene Aliases
-
UniGene
Hs.375985
Gene Chromosome Location
Chr.19: 15010727-15023271 on Build GRCh38
Transcripts
NM_173482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4669653 c.1377C>T p.N459N Chr.19: 15022996-15022996 on Build GRCh38
5611849 c.1323G>T p.K441N Chr.19: 15022942-15022942 on Build GRCh38
74025 c.1385G>A p.R462H Chr.19: 15023004-15023004 on Build GRCh38
3970758 c.1271G>T p.C424F Chr.19: 15022890-15022890 on Build GRCh38
6524697 c.1395G>A p.L465L Chr.19: 15023014-15023014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8111625 Chr.19: 15022950-15022950 on Build GRCh38
rs762791439 Chr.19: 15023000-15023007 on Build GRCh38
rs374150698 Chr.19: 15022978-15022978 on Build GRCh38
rs201842706 Chr.19: 15022951-15022951 on Build GRCh38
rs757185644 Chr.19: 15022964-15022964 on Build GRCh38
rs532537216 Chr.19: 15022997-15022997 on Build GRCh38
rs956113142 Chr.19: 15023035-15023035 on Build GRCh38
rs866222611 Chr.19: 15022930-15022930 on Build GRCh38
rs145093730 Chr.19: 15022963-15022963 on Build GRCh38
rs774648875 Chr.19: 15023049-15023049 on Build GRCh38
rs751365016 Chr.19: 15023021-15023021 on Build GRCh38
rs752355169 Chr.19: 15022949-15022949 on Build GRCh38
rs981621904 Chr.19: 15023006-15023006 on Build GRCh38
rs1016412547 Chr.19: 15022876-15022876 on Build GRCh38
rs756991217 Chr.19: 15023028-15023028 on Build GRCh38
rs10424547 Chr.19: 15022975-15022975 on Build GRCh38
rs781045152 Chr.19: 15022969-15022969 on Build GRCh38
rs17855585 Chr.19: 15022921-15022921 on Build GRCh38
rs781133010 Chr.19: 15023033-15023033 on Build GRCh38
rs764775035 Chr.19: 15022996-15022996 on Build GRCh38
rs137956196 Chr.19: 15023017-15023017 on Build GRCh38
rs753393883 Chr.19: 15022922-15022922 on Build GRCh38
rs369783890 Chr.19: 15022987-15022987 on Build GRCh38
rs754469548 Chr.19: 15022924-15022924 on Build GRCh38
rs769872174 Chr.19: 15022973-15022973 on Build GRCh38
rs772515831 Chr.19: 15023055-15023055 on Build GRCh38
rs377679495 Chr.19: 15022868-15022868 on Build GRCh38
rs749491742 Chr.19: 15022977-15022977 on Build GRCh38
rs764477337 Chr.19: 15022907-15022907 on Build GRCh38
rs774842985 Chr.19: 15022979-15022979 on Build GRCh38
rs768918482 Chr.19: 15023046-15023046 on Build GRCh38
rs934622198 Chr.19: 15022891-15022891 on Build GRCh38
rs758056018 Chr.19: 15022999-15022999 on Build GRCh38
rs763246518 Chr.19: 15022889-15022889 on Build GRCh38
rs776026795 Chr.19: 15022884-15022884 on Build GRCh38
rs745341822 Chr.19: 15022953-15022953 on Build GRCh38
rs760579083 Chr.19: 15023043-15023044 on Build GRCh38
rs769475871 Chr.19: 15022967-15022967 on Build GRCh38
rs564899590 Chr.19: 15023000-15023001 on Build GRCh38
rs140265809 Chr.19: 15022986-15022986 on Build GRCh38
rs971540411 Chr.19: 15023003-15023003 on Build GRCh38
rs749527225 Chr.19: 15023042-15023042 on Build GRCh38
rs149455236 Chr.19: 15023034-15023034 on Build GRCh38
rs756185939 Chr.19: 15023041-15023041 on Build GRCh38
rs775099299 Chr.19: 15022999-15023000 on Build GRCh38
rs745962655 Chr.19: 15022972-15022972 on Build GRCh38
rs906501792 Chr.19: 15023029-15023029 on Build GRCh38
rs775727525 Chr.19: 15022880-15022880 on Build GRCh38
rs116923487 Chr.19: 15022991-15022991 on Build GRCh38
rs151111600 Chr.19: 15022936-15022936 on Build GRCh38
rs770011391 Chr.19: 15022847-15022847 on Build GRCh38
rs375778651 Chr.19: 15023050-15023050 on Build GRCh38
rs774756657 Chr.19: 15022910-15022910 on Build GRCh38
rs914550588 Chr.19: 15023059-15023059 on Build GRCh38
rs759825779 Chr.19: 15022846-15022846 on Build GRCh38

View Full Product Details