Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99967110-99967335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZGP1
Entrez Gene ID
563
Gene Name
alpha-2-glycoprotein 1, zinc-binding
Gene Aliases
ZA2G, ZAG
UniGene
Hs.546239
Gene Chromosome Location
Chr.7: 99966727-99976112 on Build GRCh38
Transcripts
NM_001185.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5773427 c.766C>T p.H256Y Chr.7: 99967134-99967134 on Build GRCh38
6609122 c.626T>G p.V209G Chr.7: 99967274-99967274 on Build GRCh38
5763639 c.678G>A p.L226L Chr.7: 99967222-99967222 on Build GRCh38
1093915 c.751C>T p.R251W Chr.7: 99967149-99967149 on Build GRCh38
1093914 c.757G>T p.D253Y Chr.7: 99967143-99967143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377110545 Chr.7: 99967312-99967312 on Build GRCh38
rs754014053 Chr.7: 99967160-99967160 on Build GRCh38
rs771087962 Chr.7: 99967213-99967215 on Build GRCh38
rs770875487 Chr.7: 99967186-99967186 on Build GRCh38
rs879216779 Chr.7: 99967289-99967289 on Build GRCh38
rs887416437 Chr.7: 99967144-99967144 on Build GRCh38
rs768853505 Chr.7: 99967173-99967173 on Build GRCh38
rs777083732 Chr.7: 99967174-99967174 on Build GRCh38
rs762614872 Chr.7: 99967183-99967183 on Build GRCh38
rs1013692378 Chr.7: 99967187-99967187 on Build GRCh38
rs769580477 Chr.7: 99967178-99967178 on Build GRCh38
rs747199042 Chr.7: 99967188-99967188 on Build GRCh38
rs781639562 Chr.7: 99967283-99967283 on Build GRCh38
rs993986823 Chr.7: 99967236-99967236 on Build GRCh38
rs748578035 Chr.7: 99967293-99967293 on Build GRCh38
rs758931128 Chr.7: 99967170-99967170 on Build GRCh38
rs751369711 Chr.7: 99967148-99967148 on Build GRCh38
rs755510618 Chr.7: 99967278-99967278 on Build GRCh38
rs757503983 Chr.7: 99967167-99967167 on Build GRCh38
rs759542986 Chr.7: 99967212-99967212 on Build GRCh38
rs1023913171 Chr.7: 99967216-99967216 on Build GRCh38
rs780202757 Chr.7: 99967258-99967258 on Build GRCh38
rs879147313 Chr.7: 99967290-99967290 on Build GRCh38
rs112711529 Chr.7: 99967310-99967310 on Build GRCh38
rs778899781 Chr.7: 99967168-99967168 on Build GRCh38
rs758456264 Chr.7: 99967251-99967251 on Build GRCh38
rs772831043 Chr.7: 99967179-99967179 on Build GRCh38
rs767179061 Chr.7: 99967214-99967214 on Build GRCh38
rs188051315 Chr.7: 99967313-99967313 on Build GRCh38
rs752494676 Chr.7: 99967235-99967235 on Build GRCh38
rs199572012 Chr.7: 99967149-99967149 on Build GRCh38
rs143279151 Chr.7: 99967247-99967247 on Build GRCh38
rs868203395 Chr.7: 99967244-99967244 on Build GRCh38
rs769561875 Chr.7: 99967302-99967302 on Build GRCh38
rs1032762643 Chr.7: 99967280-99967280 on Build GRCh38
rs751968303 Chr.7: 99967259-99967259 on Build GRCh38
rs372045467 Chr.7: 99967196-99967196 on Build GRCh38
rs570933446 Chr.7: 99967137-99967137 on Build GRCh38
rs139788000 Chr.7: 99967171-99967171 on Build GRCh38

View Full Product Details