Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2435705-2435972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNF
Entrez Gene ID
899
Gene Name
cyclin F
Gene Aliases
FBX1, FBXO1
UniGene
Hs.1973
Gene Chromosome Location
Chr.16: 2425438-2458858 on Build GRCh38
Transcripts
NM_001323538.1 NM_001761.2 XM_017023822.1 XM_017023823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4578849 c.346C>T p.L116L Chr.16: 2435873-2435873 on Build GRCh38
1301767 c.301G>A p.E101K Chr.16: 2435828-2435828 on Build GRCh38
969031 c.300C>T p.F100F Chr.16: 2435827-2435827 on Build GRCh38
5481290 c.279-4T>A p.? Chr.16: 2435802-2435802 on Build GRCh38
3508017 c.316C>T p.L106L Chr.16: 2435843-2435843 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs575885240 Chr.16: 2435758-2435758 on Build GRCh38
rs765422854 Chr.16: 2435863-2435863 on Build GRCh38
rs775376235 Chr.16: 2435833-2435833 on Build GRCh38
rs764061587 Chr.16: 2435835-2435835 on Build GRCh38
rs140856384 Chr.16: 2435839-2435839 on Build GRCh38
rs760436368 Chr.16: 2435834-2435834 on Build GRCh38
rs770482472 Chr.16: 2435806-2435806 on Build GRCh38
rs781712247 Chr.16: 2435888-2435888 on Build GRCh38
rs754701735 Chr.16: 2435921-2435921 on Build GRCh38
rs887256259 Chr.16: 2435858-2435858 on Build GRCh38
rs12928789 Chr.16: 2435895-2435895 on Build GRCh38
rs770372021 Chr.16: 2435894-2435894 on Build GRCh38
rs780573440 Chr.16: 2435756-2435756 on Build GRCh38
rs370702853 Chr.16: 2435874-2435874 on Build GRCh38
rs746961242 Chr.16: 2435902-2435902 on Build GRCh38
rs773933025 Chr.16: 2435808-2435808 on Build GRCh38
rs753785529 Chr.16: 2435838-2435838 on Build GRCh38
rs200331327 Chr.16: 2435856-2435856 on Build GRCh38
rs968385593 Chr.16: 2435933-2435933 on Build GRCh38
rs755361178 Chr.16: 2435883-2435883 on Build GRCh38
rs554108597 Chr.16: 2435748-2435748 on Build GRCh38
rs999921804 Chr.16: 2435948-2435948 on Build GRCh38
rs748823835 Chr.16: 2435799-2435799 on Build GRCh38
rs767259178 Chr.16: 2435829-2435829 on Build GRCh38
rs931365281 Chr.16: 2435846-2435846 on Build GRCh38
rs1004814243 Chr.16: 2435873-2435873 on Build GRCh38
rs376699320 Chr.16: 2435827-2435827 on Build GRCh38
rs777040047 Chr.16: 2435783-2435783 on Build GRCh38
rs759191535 Chr.16: 2435814-2435814 on Build GRCh38
rs748610423 Chr.16: 2435891-2435891 on Build GRCh38
rs543066511 Chr.16: 2435872-2435872 on Build GRCh38
rs990719669 Chr.16: 2435843-2435843 on Build GRCh38
rs1041288772 Chr.16: 2435885-2435885 on Build GRCh38
rs144671828 Chr.16: 2435865-2435865 on Build GRCh38
rs992740598 Chr.16: 2435740-2435740 on Build GRCh38
rs1026452292 Chr.16: 2435742-2435742 on Build GRCh38
rs775054989 Chr.16: 2435903-2435903 on Build GRCh38
rs187867648 Chr.16: 2435759-2435759 on Build GRCh38
rs138778799 Chr.16: 2435881-2435881 on Build GRCh38

View Full Product Details