Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109375927-109376200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD164
Entrez Gene ID
8763
Gene Name
CD164 molecule
Gene Aliases
DFNA66, MGC-24, MUC-24, endolyn
UniGene
Hs.520313
Gene Chromosome Location
Chr.6: 109366514-109382559 on Build GRCh38
Transcripts
NM_001142401.1 NM_001142402.1 NM_001142403.1 NM_001142404.1 NM_006016.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522337 c.360C>T p.A120A Chr.6: 109376084-109376084 on Build GRCh38
6470629 c.363T>C p.N121N Chr.6: 109376081-109376081 on Build GRCh38
5522336 c.360C>T p.A120A Chr.6: 109376084-109376084 on Build GRCh38
6470630 c.363T>C p.N121N Chr.6: 109376081-109376081 on Build GRCh38
1292181 c.357A>G p.T119T Chr.6: 109376087-109376087 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749767704 Chr.6: 109376063-109376063 on Build GRCh38
rs939066643 Chr.6: 109376174-109376174 on Build GRCh38
rs771601649 Chr.6: 109376082-109376082 on Build GRCh38
rs541991303 Chr.6: 109376168-109376168 on Build GRCh38
rs1004390129 Chr.6: 109375971-109375971 on Build GRCh38
rs191085426 Chr.6: 109376049-109376049 on Build GRCh38
rs76123535 Chr.6: 109376001-109376001 on Build GRCh38
rs777765271 Chr.6: 109376062-109376062 on Build GRCh38
rs759162918 Chr.6: 109376119-109376125 on Build GRCh38
rs79147991 Chr.6: 109376089-109376089 on Build GRCh38
rs769536992 Chr.6: 109376138-109376139 on Build GRCh38
rs758135904 Chr.6: 109376032-109376032 on Build GRCh38
rs774657185 Chr.6: 109376095-109376095 on Build GRCh38
rs764887351 Chr.6: 109376145-109376145 on Build GRCh38
rs776159791 Chr.6: 109376125-109376125 on Build GRCh38
rs753360076 Chr.6: 109376143-109376143 on Build GRCh38
rs758221714 Chr.6: 109376147-109376147 on Build GRCh38
rs202232101 Chr.6: 109376148-109376148 on Build GRCh38
rs373906087 Chr.6: 109376105-109376105 on Build GRCh38
rs754389867 Chr.6: 109376179-109376179 on Build GRCh38
rs574817915 Chr.6: 109376013-109376013 on Build GRCh38
rs746881253 Chr.6: 109376046-109376046 on Build GRCh38
rs1022575756 Chr.6: 109376065-109376065 on Build GRCh38
rs775241499 Chr.6: 109376139-109376140 on Build GRCh38
rs554090332 Chr.6: 109376039-109376039 on Build GRCh38
rs761132987 Chr.6: 109376133-109376133 on Build GRCh38
rs1031677939 Chr.6: 109375980-109375980 on Build GRCh38
rs987534282 Chr.6: 109376010-109376010 on Build GRCh38
rs745564185 Chr.6: 109376061-109376061 on Build GRCh38
rs558692197 Chr.6: 109376165-109376165 on Build GRCh38
rs1019051993 Chr.6: 109376044-109376044 on Build GRCh38
rs764628680 Chr.6: 109376141-109376141 on Build GRCh38
rs773931581 Chr.6: 109376156-109376156 on Build GRCh38
rs751349352 Chr.6: 109376157-109376157 on Build GRCh38
rs771334333 Chr.6: 109376087-109376087 on Build GRCh38
rs540279572 Chr.6: 109376038-109376038 on Build GRCh38
rs369339035 Chr.6: 109376031-109376031 on Build GRCh38
rs769941614 Chr.6: 109376058-109376058 on Build GRCh38
rs759982416 Chr.6: 109376101-109376101 on Build GRCh38
rs780743213 Chr.6: 109376178-109376178 on Build GRCh38
rs756522045 Chr.6: 109376100-109376100 on Build GRCh38

View Full Product Details