Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1516001-1516252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3A
Entrez Gene ID
55210
Gene Name
ATPase family, AAA domain containing 3A
Gene Aliases
-
UniGene
Hs.23413
Gene Chromosome Location
Chr.1: 1512143-1534687 on Build GRCh38
Transcripts
NM_001170535.1 NM_001170536.1 NM_018188.3 XM_017001628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4021725 c.206-1G>T p.? Chr.1: 1516011-1516011 on Build GRCh38
3788591 c.273C>G p.S91S Chr.1: 1516079-1516079 on Build GRCh38
6410875 c.256C>A p.Q86K Chr.1: 1516062-1516062 on Build GRCh38
895136 c.269A>G p.Q90R Chr.1: 1516075-1516075 on Build GRCh38
6606403 c.220G>A p.A74T Chr.1: 1516026-1516026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368473476 Chr.1: 1516101-1516101 on Build GRCh38
rs549943434 Chr.1: 1516140-1516140 on Build GRCh38
rs576613525 Chr.1: 1516215-1516215 on Build GRCh38
rs9439459 Chr.1: 1516109-1516109 on Build GRCh38
rs771464169 Chr.1: 1516082-1516082 on Build GRCh38
rs147308420 Chr.1: 1516075-1516075 on Build GRCh38
rs567909244 Chr.1: 1516137-1516137 on Build GRCh38
rs374605918 Chr.1: 1516052-1516052 on Build GRCh38
rs2478794 Chr.1: 1516040-1516040 on Build GRCh38
rs1044660791 Chr.1: 1516150-1516150 on Build GRCh38
rs556826014 Chr.1: 1516180-1516180 on Build GRCh38
rs775596509 Chr.1: 1516020-1516020 on Build GRCh38
rs368391530 Chr.1: 1516093-1516093 on Build GRCh38
rs746496100 Chr.1: 1516126-1516126 on Build GRCh38
rs755953936 Chr.1: 1516030-1516030 on Build GRCh38
rs962048334 Chr.1: 1516158-1516159 on Build GRCh38
rs9439460 Chr.1: 1516110-1516110 on Build GRCh38
rs367565654 Chr.1: 1516109-1516110 on Build GRCh38
rs770079488 Chr.1: 1516136-1516136 on Build GRCh38
rs372999388 Chr.1: 1516183-1516183 on Build GRCh38
rs938453349 Chr.1: 1516177-1516177 on Build GRCh38
rs2478793 Chr.1: 1516141-1516141 on Build GRCh38
rs546711654 Chr.1: 1516057-1516057 on Build GRCh38
rs764695721 Chr.1: 1516051-1516051 on Build GRCh38
rs778656174 Chr.1: 1516045-1516045 on Build GRCh38
rs368069993 Chr.1: 1516118-1516118 on Build GRCh38
rs754834610 Chr.1: 1516190-1516190 on Build GRCh38
rs763026922 Chr.1: 1516108-1516108 on Build GRCh38
rs186571713 Chr.1: 1516165-1516165 on Build GRCh38
rs138594222 Chr.1: 1516035-1516035 on Build GRCh38
rs371964475 Chr.1: 1516123-1516123 on Build GRCh38
rs1004512228 Chr.1: 1516206-1516206 on Build GRCh38
rs760974299 Chr.1: 1516143-1516143 on Build GRCh38
rs996432773 Chr.1: 1516225-1516225 on Build GRCh38
rs1056827765 Chr.1: 1516182-1516182 on Build GRCh38
rs757752698 Chr.1: 1516128-1516128 on Build GRCh38
rs746249719 Chr.1: 1516133-1516133 on Build GRCh38
rs781689095 Chr.1: 1516132-1516132 on Build GRCh38
rs764981930 Chr.1: 1516119-1516119 on Build GRCh38
rs747688369 Chr.1: 1516047-1516047 on Build GRCh38
rs766313900 Chr.1: 1516111-1516111 on Build GRCh38
rs777490160 Chr.1: 1516125-1516125 on Build GRCh38
rs1015841842 Chr.1: 1516214-1516214 on Build GRCh38
rs2986577 Chr.1: 1516124-1516124 on Build GRCh38
rs754630444 Chr.1: 1516044-1516044 on Build GRCh38
rs753628565 Chr.1: 1516113-1516113 on Build GRCh38
rs757913461 Chr.1: 1516053-1516053 on Build GRCh38
rs376012620 Chr.1: 1516100-1516100 on Build GRCh38
rs576849249 Chr.1: 1516164-1516164 on Build GRCh38
rs886159202 Chr.1: 1516201-1516201 on Build GRCh38
rs750294848 Chr.1: 1516029-1516029 on Build GRCh38
rs141392786 Chr.1: 1516026-1516026 on Build GRCh38

View Full Product Details