Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2276164-2276429 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf35
Entrez Gene ID
374872
Gene Name
chromosome 19 open reading frame 35
Gene Aliases
-
UniGene
Hs.511803
Gene Chromosome Location
Chr.19: 2274632-2282182 on Build GRCh38
Transcripts
NM_198532.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6017116 c.804G>T p.E268D Chr.19: 2276298-2276298 on Build GRCh38
3531720 c.766G>A p.E256K Chr.19: 2276336-2276336 on Build GRCh38
3531721 c.740G>A p.W247* Chr.19: 2276362-2276362 on Build GRCh38
1750771 c.676C>G p.P226A Chr.19: 2276426-2276426 on Build GRCh38
6150792 c.897C>G p.G299G Chr.19: 2276205-2276205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371491982 Chr.19: 2276186-2276186 on Build GRCh38
rs777348610 Chr.19: 2276209-2276209 on Build GRCh38
rs534883075 Chr.19: 2276301-2276301 on Build GRCh38
rs943107928 Chr.19: 2276307-2276307 on Build GRCh38
rs780500874 Chr.19: 2276198-2276198 on Build GRCh38
rs748414979 Chr.19: 2276350-2276350 on Build GRCh38
rs765433092 Chr.19: 2276288-2276288 on Build GRCh38
rs368200020 Chr.19: 2276251-2276251 on Build GRCh38
rs756713943 Chr.19: 2276303-2276303 on Build GRCh38
rs867423273 Chr.19: 2276203-2276203 on Build GRCh38
rs780889442 Chr.19: 2276384-2276384 on Build GRCh38
rs752604810 Chr.19: 2276243-2276245 on Build GRCh38
rs866840806 Chr.19: 2276201-2276201 on Build GRCh38
rs771784527 Chr.19: 2276274-2276274 on Build GRCh38
rs200530567 Chr.19: 2276399-2276399 on Build GRCh38
rs374279252 Chr.19: 2276212-2276212 on Build GRCh38
rs537115116 Chr.19: 2276406-2276406 on Build GRCh38
rs775365909 Chr.19: 2276278-2276278 on Build GRCh38
rs757374508 Chr.19: 2276372-2276373 on Build GRCh38
rs764157772 Chr.19: 2276274-2276276 on Build GRCh38
rs974898244 Chr.19: 2276312-2276312 on Build GRCh38
rs745744695 Chr.19: 2276313-2276313 on Build GRCh38
rs566582464 Chr.19: 2276249-2276249 on Build GRCh38
rs774769058 Chr.19: 2276376-2276376 on Build GRCh38
rs746784189 Chr.19: 2276337-2276337 on Build GRCh38
rs759208281 Chr.19: 2276182-2276182 on Build GRCh38
rs755804524 Chr.19: 2276208-2276208 on Build GRCh38
rs765035754 Chr.19: 2276237-2276238 on Build GRCh38
rs1048932024 Chr.19: 2276324-2276324 on Build GRCh38
rs751652789 Chr.19: 2276322-2276323 on Build GRCh38
rs972972370 Chr.19: 2276247-2276247 on Build GRCh38
rs779918015 Chr.19: 2276327-2276327 on Build GRCh38
rs1040687239 Chr.19: 2276370-2276370 on Build GRCh38
rs1035824582 Chr.19: 2276227-2276227 on Build GRCh38
rs989625643 Chr.19: 2276223-2276223 on Build GRCh38
rs202010475 Chr.19: 2276341-2276341 on Build GRCh38
rs758789888 Chr.19: 2276197-2276197 on Build GRCh38
rs371752048 Chr.19: 2276380-2276380 on Build GRCh38
rs751019721 Chr.19: 2276193-2276193 on Build GRCh38
rs767802416 Chr.19: 2276383-2276383 on Build GRCh38
rs758368845 Chr.19: 2276242-2276243 on Build GRCh38
rs768914537 Chr.19: 2276282-2276282 on Build GRCh38
rs905862415 Chr.19: 2276393-2276393 on Build GRCh38
rs557687551 Chr.19: 2276372-2276372 on Build GRCh38
rs199949372 Chr.19: 2276302-2276302 on Build GRCh38
rs200177400 Chr.19: 2276338-2276338 on Build GRCh38
rs761965563 Chr.19: 2276285-2276285 on Build GRCh38
rs201170734 Chr.19: 2276367-2276367 on Build GRCh38
rs769930656 Chr.19: 2276354-2276354 on Build GRCh38
rs752225796 Chr.19: 2276204-2276204 on Build GRCh38
rs772030560 Chr.19: 2276319-2276319 on Build GRCh38
rs749926394 Chr.19: 2276400-2276400 on Build GRCh38
rs753937901 Chr.19: 2276182-2276182 on Build GRCh38
rs371731431 Chr.19: 2276297-2276297 on Build GRCh38
rs765811346 Chr.19: 2276192-2276192 on Build GRCh38
rs751895128 Chr.19: 2276233-2276233 on Build GRCh38
rs919352247 Chr.19: 2276292-2276292 on Build GRCh38
rs750922853 Chr.19: 2276290-2276290 on Build GRCh38
rs755372819 Chr.19: 2276355-2276355 on Build GRCh38
rs776761674 Chr.19: 2276284-2276284 on Build GRCh38
rs778397324 Chr.19: 2276306-2276306 on Build GRCh38
rs760440851 Chr.19: 2276280-2276280 on Build GRCh38
rs745834677 Chr.19: 2276260-2276260 on Build GRCh38
rs751940492 Chr.19: 2276299-2276299 on Build GRCh38
rs763559496 Chr.19: 2276293-2276293 on Build GRCh38
rs12609944 Chr.19: 2276236-2276236 on Build GRCh38
rs960493402 Chr.19: 2276234-2276234 on Build GRCh38
rs756913666 Chr.19: 2276396-2276396 on Build GRCh38

View Full Product Details