Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103559597-103559870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG5
Entrez Gene ID
9529
Gene Name
BCL2 associated athanogene 5
Gene Aliases
BAG-5
UniGene
Hs.5443
Gene Chromosome Location
Chr.14: 103556544-103562814 on Build GRCh38
Transcripts
NM_001015048.2 NM_001015049.2 NM_004873.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3493998 c.1314C>T p.L438L Chr.14: 103559851-103559851 on Build GRCh38
5625911 c.1420A>G p.N474D Chr.14: 103559868-103559868 on Build GRCh38
5625912 c.1297A>G p.N433D Chr.14: 103559868-103559868 on Build GRCh38
172802 c.1315G>A p.D439N Chr.14: 103559850-103559850 on Build GRCh38
3493997 c.1437C>T p.L479L Chr.14: 103559851-103559851 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571629181 Chr.14: 103559649-103559649 on Build GRCh38
rs914648111 Chr.14: 103559628-103559628 on Build GRCh38
rs567721916 Chr.14: 103559702-103559702 on Build GRCh38
rs550355142 Chr.14: 103559696-103559696 on Build GRCh38
rs775345840 Chr.14: 103559811-103559811 on Build GRCh38
rs758057337 Chr.14: 103559736-103559736 on Build GRCh38
rs753356882 Chr.14: 103559643-103559643 on Build GRCh38
rs929110915 Chr.14: 103559768-103559768 on Build GRCh38
rs1047678272 Chr.14: 103559631-103559631 on Build GRCh38
rs762995303 Chr.14: 103559822-103559822 on Build GRCh38
rs776320557 Chr.14: 103559797-103559797 on Build GRCh38
rs879790563 Chr.14: 103559647-103559647 on Build GRCh38
rs768533731 Chr.14: 103559775-103559775 on Build GRCh38
rs907697014 Chr.14: 103559652-103559654 on Build GRCh38
rs759647800 Chr.14: 103559697-103559697 on Build GRCh38
rs941898619 Chr.14: 103559653-103559653 on Build GRCh38
rs536452354 Chr.14: 103559735-103559735 on Build GRCh38
rs944632467 Chr.14: 103559732-103559732 on Build GRCh38
rs567866975 Chr.14: 103559776-103559776 on Build GRCh38
rs1012629870 Chr.14: 103559839-103559839 on Build GRCh38
rs756762907 Chr.14: 103559659-103559659 on Build GRCh38
rs995814594 Chr.14: 103559708-103559708 on Build GRCh38
rs770596839 Chr.14: 103559787-103559787 on Build GRCh38
rs190873312 Chr.14: 103559745-103559745 on Build GRCh38
rs779758720 Chr.14: 103559769-103559769 on Build GRCh38
rs777272419 Chr.14: 103559779-103559779 on Build GRCh38
rs750080988 Chr.14: 103559727-103559727 on Build GRCh38
rs370742587 Chr.14: 103559808-103559808 on Build GRCh38
rs759285906 Chr.14: 103559805-103559805 on Build GRCh38
rs746486280 Chr.14: 103559780-103559780 on Build GRCh38
rs985865923 Chr.14: 103559711-103559711 on Build GRCh38
rs368544168 Chr.14: 103559774-103559774 on Build GRCh38
rs887523894 Chr.14: 103559793-103559793 on Build GRCh38
rs553110823 Chr.14: 103559816-103559816 on Build GRCh38
rs774510576 Chr.14: 103559693-103559693 on Build GRCh38
rs75414319 Chr.14: 103559767-103559767 on Build GRCh38
rs1037382421 Chr.14: 103559656-103559656 on Build GRCh38

View Full Product Details