Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169804020-169804279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf112
Entrez Gene ID
55732
Gene Name
chromosome 1 open reading frame 112
Gene Aliases
-
UniGene
Hs.443551
Gene Chromosome Location
Chr.1: 169794730-169854080 on Build GRCh38
Transcripts
NM_001320047.1 NM_001320048.1 NM_001320050.1 NM_001320051.1 NM_018186.3 XM_005245317.4 XM_011509735.2 XM_017001722.1 XM_017001723.1 XM_017001724.1 XM_017001725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7417015 c.640A>G p.N214D Chr.1: 169804228-169804228 on Build GRCh38
463383 c.445G>C p.D149H Chr.1: 169804207-169804207 on Build GRCh38
5824749 c.384C>G p.A128A Chr.1: 169804146-169804146 on Build GRCh38
7306585 c.632A>T p.D211V Chr.1: 169804220-169804220 on Build GRCh38
6354209 c.588G>A p.L196L Chr.1: 169804176-169804176 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995008010 Chr.1: 169804185-169804185 on Build GRCh38
rs767098070 Chr.1: 169804206-169804206 on Build GRCh38
rs374141112 Chr.1: 169804193-169804193 on Build GRCh38
rs960385281 Chr.1: 169804238-169804238 on Build GRCh38
rs771325590 Chr.1: 169804171-169804171 on Build GRCh38
rs748257437 Chr.1: 169804158-169804158 on Build GRCh38
rs747257312 Chr.1: 169804170-169804170 on Build GRCh38
rs199744208 Chr.1: 169804196-169804196 on Build GRCh38
rs749241377 Chr.1: 169804108-169804108 on Build GRCh38
rs779111872 Chr.1: 169804148-169804148 on Build GRCh38
rs1053013445 Chr.1: 169804052-169804052 on Build GRCh38
rs1027443969 Chr.1: 169804188-169804188 on Build GRCh38
rs144946376 Chr.1: 169804190-169804190 on Build GRCh38
rs754823645 Chr.1: 169804213-169804215 on Build GRCh38
rs773584153 Chr.1: 169804165-169804165 on Build GRCh38
rs751393015 Chr.1: 169804197-169804199 on Build GRCh38
rs760067677 Chr.1: 169804182-169804182 on Build GRCh38
rs11590604 Chr.1: 169804176-169804176 on Build GRCh38
rs373392728 Chr.1: 169804067-169804067 on Build GRCh38
rs34578849 Chr.1: 169804074-169804075 on Build GRCh38
rs370208533 Chr.1: 169804103-169804103 on Build GRCh38
rs780043952 Chr.1: 169804195-169804196 on Build GRCh38
rs755784616 Chr.1: 169804213-169804213 on Build GRCh38
rs966823176 Chr.1: 169804072-169804072 on Build GRCh38
rs772356317 Chr.1: 169804161-169804161 on Build GRCh38
rs143006454 Chr.1: 169804210-169804210 on Build GRCh38
rs755950138 Chr.1: 169804095-169804095 on Build GRCh38
rs201886828 Chr.1: 169804085-169804085 on Build GRCh38
rs534717429 Chr.1: 169804198-169804198 on Build GRCh38
rs367824430 Chr.1: 169804233-169804233 on Build GRCh38
rs199703302 Chr.1: 169804069-169804069 on Build GRCh38
rs190533081 Chr.1: 169804066-169804066 on Build GRCh38
rs756856339 Chr.1: 169804076-169804076 on Build GRCh38
rs201023910 Chr.1: 169804080-169804080 on Build GRCh38
rs765757408 Chr.1: 169804183-169804183 on Build GRCh38
rs1038951222 Chr.1: 169804053-169804053 on Build GRCh38
rs769220936 Chr.1: 169804079-169804079 on Build GRCh38
rs147711417 Chr.1: 169804089-169804089 on Build GRCh38

View Full Product Details