Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11550100-11550371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLK
Entrez Gene ID
640
Gene Name
BLK proto-oncogene, Src family tyrosine kinase
Gene Aliases
MODY11
UniGene
Hs.146591
Gene Chromosome Location
Chr.8: 11493991-11564599 on Build GRCh38
Transcripts
NM_001715.2 XM_011543824.1 XM_011543825.2 XM_011543827.1 XM_011543828.2 XM_011543829.2 XM_017013730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5989875 c.391C>T p.R131W Chr.8: 11550181-11550181 on Build GRCh38
1686045 c.440C>T p.S147F Chr.8: 11550230-11550230 on Build GRCh38
1184745 c.412C>T p.L138F Chr.8: 11550202-11550202 on Build GRCh38
7080029 c.431_432insGGCCG p.S147fs*20 Chr.8: 11550221-11550222 on Build GRCh38
5901535 c.373T>C p.F125L Chr.8: 11550163-11550163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753053621 Chr.8: 11550125-11550125 on Build GRCh38
rs773415274 Chr.8: 11550303-11550303 on Build GRCh38
rs746568651 Chr.8: 11550268-11550268 on Build GRCh38
rs1028964004 Chr.8: 11550171-11550171 on Build GRCh38
rs547792358 Chr.8: 11550254-11550254 on Build GRCh38
rs777465313 Chr.8: 11550226-11550226 on Build GRCh38
rs767696277 Chr.8: 11550301-11550301 on Build GRCh38
rs199898752 Chr.8: 11550295-11550295 on Build GRCh38
rs569437248 Chr.8: 11550124-11550124 on Build GRCh38
rs758030935 Chr.8: 11550193-11550194 on Build GRCh38
rs939336015 Chr.8: 11550201-11550201 on Build GRCh38
rs766430463 Chr.8: 11550307-11550307 on Build GRCh38
rs547109926 Chr.8: 11550314-11550314 on Build GRCh38
rs750524454 Chr.8: 11550227-11550227 on Build GRCh38
rs956730553 Chr.8: 11550189-11550189 on Build GRCh38
rs371289373 Chr.8: 11550309-11550309 on Build GRCh38
rs751094359 Chr.8: 11550241-11550242 on Build GRCh38
rs749767816 Chr.8: 11550290-11550290 on Build GRCh38
rs562247916 Chr.8: 11550214-11550214 on Build GRCh38
rs202149014 Chr.8: 11550226-11550226 on Build GRCh38
rs368987710 Chr.8: 11550267-11550267 on Build GRCh38
rs776062791 Chr.8: 11550187-11550187 on Build GRCh38
rs749146255 Chr.8: 11550156-11550156 on Build GRCh38
rs137986278 Chr.8: 11550157-11550157 on Build GRCh38
rs73663163 Chr.8: 11550181-11550181 on Build GRCh38
rs756212051 Chr.8: 11550237-11550237 on Build GRCh38
rs751857293 Chr.8: 11550223-11550223 on Build GRCh38
rs371553190 Chr.8: 11550306-11550306 on Build GRCh38
rs762219322 Chr.8: 11550299-11550299 on Build GRCh38
rs145058498 Chr.8: 11550274-11550274 on Build GRCh38
rs1037993231 Chr.8: 11550327-11550327 on Build GRCh38
rs534542140 Chr.8: 11550313-11550313 on Build GRCh38
rs200916733 Chr.8: 11550239-11550239 on Build GRCh38
rs371418729 Chr.8: 11550282-11550282 on Build GRCh38
rs1056884367 Chr.8: 11550217-11550217 on Build GRCh38
rs771805021 Chr.8: 11550330-11550330 on Build GRCh38
rs751765814 Chr.8: 11550129-11550129 on Build GRCh38
rs7836533 Chr.8: 11550225-11550225 on Build GRCh38
rs557414333 Chr.8: 11550347-11550347 on Build GRCh38
rs866807537 Chr.8: 11550173-11550173 on Build GRCh38
rs375035401 Chr.8: 11550255-11550255 on Build GRCh38
rs754947956 Chr.8: 11550312-11550312 on Build GRCh38
rs139003574 Chr.8: 11550186-11550186 on Build GRCh38
rs764499421 Chr.8: 11550220-11550220 on Build GRCh38
rs963412427 Chr.8: 11550154-11550154 on Build GRCh38
rs144615291 Chr.8: 11550182-11550182 on Build GRCh38
rs757428982 Chr.8: 11550130-11550130 on Build GRCh38
rs756761426 Chr.8: 11550206-11550206 on Build GRCh38
rs781087154 Chr.8: 11550131-11550131 on Build GRCh38
rs370810777 Chr.8: 11550150-11550150 on Build GRCh38
rs769179080 Chr.8: 11550293-11550293 on Build GRCh38
rs954818860 Chr.8: 11550178-11550178 on Build GRCh38
rs750408108 Chr.8: 11550149-11550149 on Build GRCh38
rs190456319 Chr.8: 11550315-11550315 on Build GRCh38
rs192242424 Chr.8: 11550122-11550122 on Build GRCh38
rs535804924 Chr.8: 11550329-11550329 on Build GRCh38
rs546054120 Chr.8: 11550161-11550161 on Build GRCh38
rs185831709 Chr.8: 11550244-11550244 on Build GRCh38

View Full Product Details