Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 75234817-75235090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAR1
Entrez Gene ID
9564
Gene Name
BCAR1, Cas family scaffolding protein
Gene Aliases
CAS, CAS1, CASS1, CRKAS, P130Cas
UniGene
Hs.479747
Gene Chromosome Location
Chr.16: 75229030-75268053 on Build GRCh38
Transcripts
NM_001170714.1 NM_001170715.1 NM_001170716.1 NM_001170717.1 NM_001170718.1 NM_001170719.1 NM_001170720.1 NM_001170721.1 NM_014567.3 XM_005256258.4 XM_005256259.4 XM_005256260.4 XM_011523475.2 XM_017023897.1 XM_017023898.1 XM_017023899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5363437 c.1893G>A p.P631P Chr.16: 75235060-75235060 on Build GRCh38
6564558 c.1900G>A p.E634K Chr.16: 75235053-75235053 on Build GRCh38
6998694 c.2020A>G p.K674E Chr.16: 75235017-75235017 on Build GRCh38
6998698 c.1882A>G p.K628E Chr.16: 75235017-75235017 on Build GRCh38
6560682 c.2086G>A p.D696N Chr.16: 75234951-75234951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767203752 Chr.16: 75234932-75234932 on Build GRCh38
rs904667889 Chr.16: 75234869-75234869 on Build GRCh38
rs762737735 Chr.16: 75234993-75234993 on Build GRCh38
rs764627155 Chr.16: 75234855-75234855 on Build GRCh38
rs368429444 Chr.16: 75234919-75234919 on Build GRCh38
rs759268877 Chr.16: 75234852-75234852 on Build GRCh38
rs749449513 Chr.16: 75234875-75234875 on Build GRCh38
rs953220414 Chr.16: 75235049-75235049 on Build GRCh38
rs1014503513 Chr.16: 75234990-75234990 on Build GRCh38
rs767423879 Chr.16: 75235022-75235022 on Build GRCh38
rs746174916 Chr.16: 75234969-75234969 on Build GRCh38
rs774063676 Chr.16: 75235009-75235009 on Build GRCh38
rs375884665 Chr.16: 75234931-75234931 on Build GRCh38
rs768493127 Chr.16: 75235057-75235057 on Build GRCh38
rs1053124126 Chr.16: 75234939-75234939 on Build GRCh38
rs770252990 Chr.16: 75235051-75235051 on Build GRCh38
rs773712119 Chr.16: 75234847-75234847 on Build GRCh38
rs766189830 Chr.16: 75235028-75235028 on Build GRCh38
rs371886799 Chr.16: 75234872-75234872 on Build GRCh38
rs925471936 Chr.16: 75234906-75234906 on Build GRCh38
rs753690396 Chr.16: 75235068-75235068 on Build GRCh38
rs139515923 Chr.16: 75234955-75234955 on Build GRCh38
rs775246016 Chr.16: 75234981-75234981 on Build GRCh38
rs534427070 Chr.16: 75235062-75235062 on Build GRCh38
rs763732321 Chr.16: 75234928-75234928 on Build GRCh38
rs781681087 Chr.16: 75235047-75235047 on Build GRCh38
rs758664765 Chr.16: 75235035-75235035 on Build GRCh38
rs747447339 Chr.16: 75234957-75234957 on Build GRCh38
rs779047175 Chr.16: 75234950-75234950 on Build GRCh38
rs375056497 Chr.16: 75234879-75234879 on Build GRCh38
rs546382267 Chr.16: 75234925-75234925 on Build GRCh38
rs777098295 Chr.16: 75234913-75234913 on Build GRCh38
rs376744472 Chr.16: 75235060-75235060 on Build GRCh38
rs771169399 Chr.16: 75234908-75234908 on Build GRCh38
rs751064514 Chr.16: 75234858-75234860 on Build GRCh38
rs753392680 Chr.16: 75234850-75234850 on Build GRCh38
rs780614503 Chr.16: 75235052-75235052 on Build GRCh38
rs752310861 Chr.16: 75234863-75234863 on Build GRCh38
rs755868116 Chr.16: 75234943-75234943 on Build GRCh38
rs748939995 Chr.16: 75235055-75235055 on Build GRCh38
rs368776894 Chr.16: 75234953-75234953 on Build GRCh38
rs751573552 Chr.16: 75234867-75234867 on Build GRCh38
rs201994321 Chr.16: 75234848-75234848 on Build GRCh38
rs914192217 Chr.16: 75234940-75234940 on Build GRCh38
rs529896838 Chr.16: 75234865-75234865 on Build GRCh38
rs61736821 Chr.16: 75235025-75235025 on Build GRCh38
rs777798612 Chr.16: 75234866-75234866 on Build GRCh38
rs112019183 Chr.16: 75235064-75235064 on Build GRCh38
rs371754964 Chr.16: 75235011-75235011 on Build GRCh38
rs776851986 Chr.16: 75234964-75234964 on Build GRCh38
rs576248634 Chr.16: 75234962-75234962 on Build GRCh38
rs568726629 Chr.16: 75235061-75235061 on Build GRCh38
rs772150390 Chr.16: 75234882-75234882 on Build GRCh38
rs199953279 Chr.16: 75234849-75234849 on Build GRCh38
rs911809541 Chr.16: 75235063-75235063 on Build GRCh38
rs747179179 Chr.16: 75235039-75235039 on Build GRCh38
rs752881055 Chr.16: 75235032-75235032 on Build GRCh38
rs199700159 Chr.16: 75235046-75235046 on Build GRCh38
rs747581427 Chr.16: 75234890-75234890 on Build GRCh38
rs763679369 Chr.16: 75235004-75235004 on Build GRCh38
rs368873588 Chr.16: 75235065-75235065 on Build GRCh38
rs200359903 Chr.16: 75235041-75235041 on Build GRCh38
rs773659192 Chr.16: 75234889-75234889 on Build GRCh38
rs961694765 Chr.16: 75234995-75234995 on Build GRCh38
rs750299982 Chr.16: 75234941-75234941 on Build GRCh38
rs201830527 Chr.16: 75234927-75234927 on Build GRCh38
rs199823639 Chr.16: 75235048-75235048 on Build GRCh38
rs771523501 Chr.16: 75234960-75234960 on Build GRCh38
rs748162108 Chr.16: 75234881-75234881 on Build GRCh38
rs372478343 Chr.16: 75234870-75234870 on Build GRCh38
rs778961324 Chr.16: 75234880-75234880 on Build GRCh38
rs770307875 Chr.16: 75234978-75234978 on Build GRCh38
rs748391002 Chr.16: 75234951-75234951 on Build GRCh38

View Full Product Details