Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21074668-21074905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF40
Entrez Gene ID
55701
Gene Name
Rho guanine nucleotide exchange factor 40
Gene Aliases
SOLO
UniGene
Hs.35125
Gene Chromosome Location
Chr.14: 21061276-21090248 on Build GRCh38
Transcripts
NM_001278529.1 NM_001278530.1 NM_018071.4 XM_005267844.2 XM_011536937.2 XM_011536938.2 XM_017021434.1 XM_017021435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7356362 c.1151G>A p.R384Q Chr.14: 21074881-21074881 on Build GRCh38
954345 c.1067_1069delGAG p.G362delG Chr.14: 21074797-21074799 on Build GRCh38
2029510 c.1089delG p.A365fs*36 Chr.14: 21074819-21074819 on Build GRCh38
5500574 c.939A>G p.G313G Chr.14: 21074669-21074669 on Build GRCh38
6317252 c.1018G>T p.A340S Chr.14: 21074748-21074748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201422774 Chr.14: 21074843-21074843 on Build GRCh38
rs752752530 Chr.14: 21074728-21074728 on Build GRCh38
rs779118561 Chr.14: 21074751-21074751 on Build GRCh38
rs756950029 Chr.14: 21074747-21074747 on Build GRCh38
rs368911475 Chr.14: 21074690-21074690 on Build GRCh38
rs773704607 Chr.14: 21074886-21074886 on Build GRCh38
rs545146973 Chr.14: 21074800-21074802 on Build GRCh38
rs778668757 Chr.14: 21074847-21074847 on Build GRCh38
rs566211084 Chr.14: 21074766-21074766 on Build GRCh38
rs776964288 Chr.14: 21074786-21074786 on Build GRCh38
rs769175953 Chr.14: 21074781-21074781 on Build GRCh38
rs756068727 Chr.14: 21074736-21074736 on Build GRCh38
rs772205232 Chr.14: 21074761-21074761 on Build GRCh38
rs746021583 Chr.14: 21074752-21074752 on Build GRCh38
rs371889536 Chr.14: 21074697-21074697 on Build GRCh38
rs749135684 Chr.14: 21074738-21074738 on Build GRCh38
rs555879231 Chr.14: 21074856-21074856 on Build GRCh38
rs766088609 Chr.14: 21074701-21074701 on Build GRCh38
rs116108291 Chr.14: 21074803-21074803 on Build GRCh38
rs1054121927 Chr.14: 21074858-21074858 on Build GRCh38
rs767198389 Chr.14: 21074799-21074799 on Build GRCh38
rs749290845 Chr.14: 21074797-21074799 on Build GRCh38
rs769911992 Chr.14: 21074735-21074736 on Build GRCh38
rs771192751 Chr.14: 21074796-21074797 on Build GRCh38
rs751209427 Chr.14: 21074705-21074705 on Build GRCh38
rs371942296 Chr.14: 21074708-21074708 on Build GRCh38
rs781723884 Chr.14: 21074879-21074879 on Build GRCh38
rs780157076 Chr.14: 21074861-21074861 on Build GRCh38
rs755535404 Chr.14: 21074838-21074838 on Build GRCh38
rs1046089901 Chr.14: 21074777-21074777 on Build GRCh38
rs764111899 Chr.14: 21074839-21074839 on Build GRCh38
rs777482871 Chr.14: 21074737-21074737 on Build GRCh38
rs151315853 Chr.14: 21074873-21074873 on Build GRCh38
rs757090653 Chr.14: 21074844-21074844 on Build GRCh38
rs202017004 Chr.14: 21074848-21074848 on Build GRCh38
rs762262218 Chr.14: 21074790-21074790 on Build GRCh38
rs140691233 Chr.14: 21074792-21074792 on Build GRCh38
rs759260312 Chr.14: 21074793-21074793 on Build GRCh38
rs949886572 Chr.14: 21074715-21074715 on Build GRCh38
rs140605954 Chr.14: 21074881-21074881 on Build GRCh38
rs775533079 Chr.14: 21074765-21074765 on Build GRCh38
rs377320500 Chr.14: 21074797-21074802 on Build GRCh38
rs767071823 Chr.14: 21074725-21074725 on Build GRCh38
rs747134208 Chr.14: 21074869-21074869 on Build GRCh38

View Full Product Details