Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74078119-74078366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD3
Entrez Gene ID
26005
Gene Name
C2 calcium dependent domain containing 3
Gene Aliases
OFD14
UniGene
Hs.557938
Gene Chromosome Location
Chr.11: 74012578-74171210 on Build GRCh38
Transcripts
NM_001286577.1 NM_015531.5 XM_011544904.1 XM_011544905.1 XM_017017510.1 XM_017017511.1 XM_017017512.1 XM_017017513.1 XM_017017514.1 XM_017017515.1 XM_017017516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2017325 c.4436delC p.P1479fs*12 Chr.11: 74078282-74078282 on Build GRCh38
4971726 c.4356G>A p.K1452K Chr.11: 74078362-74078362 on Build GRCh38
279365 c.4356G>T p.K1452N Chr.11: 74078362-74078362 on Build GRCh38
2017321 c.4457G>T p.R1486M Chr.11: 74078261-74078261 on Build GRCh38
6482112 c.4476C>A p.I1492I Chr.11: 74078242-74078242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752449834 Chr.11: 74078165-74078165 on Build GRCh38
rs769163998 Chr.11: 74078211-74078211 on Build GRCh38
rs753690815 Chr.11: 74078146-74078146 on Build GRCh38
rs750621218 Chr.11: 74078300-74078300 on Build GRCh38
rs201031035 Chr.11: 74078163-74078163 on Build GRCh38
rs770327685 Chr.11: 74078248-74078248 on Build GRCh38
rs755449364 Chr.11: 74078330-74078330 on Build GRCh38
rs759426506 Chr.11: 74078149-74078149 on Build GRCh38
rs529606285 Chr.11: 74078291-74078291 on Build GRCh38
rs113074900 Chr.11: 74078219-74078219 on Build GRCh38
rs75137555 Chr.11: 74078287-74078287 on Build GRCh38
rs202159207 Chr.11: 74078270-74078270 on Build GRCh38
rs978918692 Chr.11: 74078282-74078282 on Build GRCh38
rs145579245 Chr.11: 74078228-74078228 on Build GRCh38
rs761331220 Chr.11: 74078324-74078344 on Build GRCh38
rs753886796 Chr.11: 74078329-74078329 on Build GRCh38
rs372607355 Chr.11: 74078207-74078207 on Build GRCh38
rs1031159988 Chr.11: 74078252-74078252 on Build GRCh38
rs776160943 Chr.11: 74078267-74078267 on Build GRCh38
rs926095562 Chr.11: 74078283-74078283 on Build GRCh38
rs756114694 Chr.11: 74078189-74078189 on Build GRCh38
rs766398654 Chr.11: 74078321-74078321 on Build GRCh38
rs772828345 Chr.11: 74078279-74078279 on Build GRCh38
rs757475399 Chr.11: 74078171-74078171 on Build GRCh38
rs750568043 Chr.11: 74078185-74078185 on Build GRCh38
rs147639769 Chr.11: 74078182-74078182 on Build GRCh38
rs541467149 Chr.11: 74078257-74078257 on Build GRCh38
rs760305311 Chr.11: 74078240-74078240 on Build GRCh38
rs1039952638 Chr.11: 74078214-74078214 on Build GRCh38
rs765157060 Chr.11: 74078281-74078281 on Build GRCh38
rs764966083 Chr.11: 74078150-74078150 on Build GRCh38
rs756308294 Chr.11: 74078304-74078304 on Build GRCh38
rs748461924 Chr.11: 74078224-74078224 on Build GRCh38
rs946242805 Chr.11: 74078264-74078264 on Build GRCh38
rs1007159030 Chr.11: 74078188-74078188 on Build GRCh38
rs772998974 Chr.11: 74078231-74078231 on Build GRCh38
rs780658413 Chr.11: 74078192-74078192 on Build GRCh38
rs1048880011 Chr.11: 74078168-74078168 on Build GRCh38

View Full Product Details