Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35462317-35462590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP250
Entrez Gene ID
11190
Gene Name
centrosomal protein 250
Gene Aliases
C-NAP1, CEP2, CNAP1
UniGene
Hs.443976
Gene Chromosome Location
Chr.20: 35455138-35517531 on Build GRCh38
Transcripts
NM_001318219.1 NM_007186.5 XM_005260262.4 XM_005260263.4 XM_005260264.4 XM_006723690.3 XM_006723691.1 XM_006723692.3 XM_006723693.3 XM_006723694.3 XM_011528517.2 XM_011528518.2 XM_017027617.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
402208 c.178C>A p.Q60K Chr.20: 35462545-35462545 on Build GRCh38
4136848 c.108A>C p.A36A Chr.20: 35462475-35462475 on Build GRCh38
77458 c.139G>A p.E47K Chr.20: 35462506-35462506 on Build GRCh38
4515848 c.134_135CC>TT p.S45F Chr.20: 35462501-35462502 on Build GRCh38
6585461 c.4G>C p.E2Q Chr.20: 35462371-35462371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753936148 Chr.20: 35462542-35462542 on Build GRCh38
rs776165342 Chr.20: 35462479-35462479 on Build GRCh38
rs780133224 Chr.20: 35462344-35462344 on Build GRCh38
rs377496493 Chr.20: 35462342-35462342 on Build GRCh38
rs772742308 Chr.20: 35462440-35462440 on Build GRCh38
rs770366322 Chr.20: 35462475-35462475 on Build GRCh38
rs978101644 Chr.20: 35462568-35462568 on Build GRCh38
rs755659951 Chr.20: 35462394-35462394 on Build GRCh38
rs959582492 Chr.20: 35462437-35462437 on Build GRCh38
rs778903424 Chr.20: 35462373-35462373 on Build GRCh38
rs1049017165 Chr.20: 35462545-35462545 on Build GRCh38
rs747946549 Chr.20: 35462397-35462397 on Build GRCh38
rs749217685 Chr.20: 35462362-35462362 on Build GRCh38
rs756320467 Chr.20: 35462521-35462521 on Build GRCh38
rs755026501 Chr.20: 35462548-35462548 on Build GRCh38
rs752611723 Chr.20: 35462569-35462569 on Build GRCh38
rs766413199 Chr.20: 35462537-35462537 on Build GRCh38
rs779765417 Chr.20: 35462454-35462454 on Build GRCh38
rs966662221 Chr.20: 35462566-35462566 on Build GRCh38
rs774753773 Chr.20: 35462501-35462501 on Build GRCh38
rs375512982 Chr.20: 35462485-35462485 on Build GRCh38
rs995672495 Chr.20: 35462540-35462540 on Build GRCh38
rs767867819 Chr.20: 35462509-35462509 on Build GRCh38
rs1001295483 Chr.20: 35462374-35462374 on Build GRCh38
rs778993326 Chr.20: 35462563-35462563 on Build GRCh38
rs762170976 Chr.20: 35462502-35462502 on Build GRCh38
rs747124216 Chr.20: 35462363-35462363 on Build GRCh38
rs200667700 Chr.20: 35462369-35462369 on Build GRCh38
rs547906210 Chr.20: 35462486-35462486 on Build GRCh38
rs141393541 Chr.20: 35462416-35462416 on Build GRCh38
rs750633022 Chr.20: 35462510-35462510 on Build GRCh38
rs746610415 Chr.20: 35462458-35462458 on Build GRCh38

View Full Product Details