Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75104140-75104401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD31
Entrez Gene ID
256006
Gene Name
ankyrin repeat domain 31
Gene Aliases
-
UniGene
Hs.482533
Gene Chromosome Location
Chr.5: 75068297-75236897 on Build GRCh38
Transcripts
NM_001164443.1 XM_011543298.2 XM_011543299.1 XM_011543300.2 XM_011543301.2 XM_011543302.2 XM_017009317.1 XM_017009318.1 XM_017009319.1 XM_017009320.1 XM_017009321.1

SNP Detail

SNP ID Chromosome Location
rs908065659 Chr.5: 75104273-75104273 on Build GRCh38
rs982229410 Chr.5: 75104204-75104204 on Build GRCh38
rs375387931 Chr.5: 75104357-75104357 on Build GRCh38
rs986072827 Chr.5: 75104369-75104369 on Build GRCh38
rs748973509 Chr.5: 75104380-75104380 on Build GRCh38
rs1035129855 Chr.5: 75104324-75104324 on Build GRCh38
rs758339025 Chr.5: 75104208-75104208 on Build GRCh38
rs79702707 Chr.5: 75104326-75104326 on Build GRCh38
rs923353155 Chr.5: 75104359-75104359 on Build GRCh38
rs769331261 Chr.5: 75104345-75104345 on Build GRCh38
rs746746627 Chr.5: 75104317-75104317 on Build GRCh38
rs745414673 Chr.5: 75104335-75104335 on Build GRCh38
rs374127019 Chr.5: 75104372-75104372 on Build GRCh38
rs777717221 Chr.5: 75104282-75104282 on Build GRCh38
rs931992078 Chr.5: 75104370-75104370 on Build GRCh38
rs140530028 Chr.5: 75104185-75104185 on Build GRCh38
rs985035882 Chr.5: 75104201-75104201 on Build GRCh38
rs761768750 Chr.5: 75104328-75104328 on Build GRCh38
rs760988894 Chr.5: 75104211-75104211 on Build GRCh38

View Full Product Details