Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27549146-27549389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHDC1
Entrez Gene ID
27245
Gene Name
AT-hook DNA binding motif containing 1
Gene Aliases
MRD25
UniGene
Hs.469280
Gene Chromosome Location
Chr.1: 27534245-27604178 on Build GRCh38
Transcripts
NM_001029882.3 XM_005245848.3 XM_005245849.3 XM_005245850.3 XM_005245851.3 XM_005245852.3 XM_011541255.1 XM_011541256.2 XM_011541257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6596636 c.2943C>T p.F981F Chr.1: 27549173-27549173 on Build GRCh38
5806060 c.2816G>T p.R939L Chr.1: 27549300-27549300 on Build GRCh38
5053365 c.2917G>A p.G973S Chr.1: 27549199-27549199 on Build GRCh38
6312631 c.2916C>A p.G972G Chr.1: 27549200-27549200 on Build GRCh38
1687282 c.2759C>T p.T920I Chr.1: 27549357-27549357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748411152 Chr.1: 27549207-27549207 on Build GRCh38
rs4908364 Chr.1: 27549313-27549313 on Build GRCh38
rs776537183 Chr.1: 27549320-27549320 on Build GRCh38
rs759357518 Chr.1: 27549230-27549230 on Build GRCh38
rs752125312 Chr.1: 27549277-27549277 on Build GRCh38
rs747339496 Chr.1: 27549202-27549202 on Build GRCh38
rs752525457 Chr.1: 27549172-27549172 on Build GRCh38
rs758990586 Chr.1: 27549191-27549191 on Build GRCh38
rs760400267 Chr.1: 27549249-27549249 on Build GRCh38
rs748308045 Chr.1: 27549288-27549288 on Build GRCh38
rs199652856 Chr.1: 27549366-27549366 on Build GRCh38
rs780536761 Chr.1: 27549194-27549194 on Build GRCh38
rs951049067 Chr.1: 27549227-27549227 on Build GRCh38
rs369475345 Chr.1: 27549293-27549293 on Build GRCh38
rs753427636 Chr.1: 27549263-27549263 on Build GRCh38
rs753538351 Chr.1: 27549178-27549178 on Build GRCh38
rs773496105 Chr.1: 27549359-27549359 on Build GRCh38
rs777736953 Chr.1: 27549343-27549343 on Build GRCh38
rs761339410 Chr.1: 27549322-27549322 on Build GRCh38
rs763881099 Chr.1: 27549261-27549261 on Build GRCh38
rs563439857 Chr.1: 27549301-27549301 on Build GRCh38
rs778551423 Chr.1: 27549187-27549187 on Build GRCh38
rs758014017 Chr.1: 27549268-27549268 on Build GRCh38
rs776936156 Chr.1: 27549203-27549203 on Build GRCh38
rs375179294 Chr.1: 27549267-27549267 on Build GRCh38
rs746971119 Chr.1: 27549365-27549365 on Build GRCh38
rs200249799 Chr.1: 27549183-27549183 on Build GRCh38
rs587779767 Chr.1: 27549218-27549218 on Build GRCh38
rs767288243 Chr.1: 27549171-27549171 on Build GRCh38
rs775334923 Chr.1: 27549307-27549307 on Build GRCh38
rs760672095 Chr.1: 27549308-27549308 on Build GRCh38
rs373401870 Chr.1: 27549368-27549368 on Build GRCh38
rs1014044758 Chr.1: 27549218-27549218 on Build GRCh38
rs150289747 Chr.1: 27549289-27549289 on Build GRCh38
rs764905350 Chr.1: 27549323-27549323 on Build GRCh38
rs781734351 Chr.1: 27549283-27549283 on Build GRCh38
rs950851633 Chr.1: 27549234-27549234 on Build GRCh38
rs749511928 Chr.1: 27549367-27549367 on Build GRCh38
rs151218177 Chr.1: 27549266-27549266 on Build GRCh38
rs749337559 Chr.1: 27549300-27549300 on Build GRCh38
rs983774136 Chr.1: 27549237-27549237 on Build GRCh38
rs533069538 Chr.1: 27549231-27549231 on Build GRCh38
rs773401303 Chr.1: 27549226-27549226 on Build GRCh38
rs750067278 Chr.1: 27549330-27549330 on Build GRCh38
rs886041781 Chr.1: 27549208-27549208 on Build GRCh38
rs762495070 Chr.1: 27549338-27549338 on Build GRCh38
rs752112717 Chr.1: 27549347-27549347 on Build GRCh38
rs909478940 Chr.1: 27549239-27549239 on Build GRCh38
rs200837232 Chr.1: 27549353-27549353 on Build GRCh38
rs780659388 Chr.1: 27549271-27549271 on Build GRCh38
rs565666869 Chr.1: 27549364-27549364 on Build GRCh38
rs775424751 Chr.1: 27549232-27549232 on Build GRCh38
rs555542437 Chr.1: 27549173-27549173 on Build GRCh38
rs769762214 Chr.1: 27549217-27549217 on Build GRCh38
rs142275561 Chr.1: 27549188-27549188 on Build GRCh38
rs112729214 Chr.1: 27549176-27549176 on Build GRCh38

View Full Product Details