Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240891753-240892024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf54
Entrez Gene ID
79919
Gene Name
chromosome 2 open reading frame 54
Gene Aliases
-
UniGene
Hs.193745
Gene Chromosome Location
Chr.2: 240886048-240896889 on Build GRCh38
Transcripts
NM_001085437.2 NM_001282921.1 NM_024861.3 XM_011511877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7133443 c.71C>T p.S24L Chr.2: 240891760-240891760 on Build GRCh38
5491616 c.43T>C p.W15R Chr.2: 240891897-240891897 on Build GRCh38
7133442 c.518C>T p.S173L Chr.2: 240891760-240891760 on Build GRCh38
3766555 c.61G>C p.A21P Chr.2: 240891879-240891879 on Build GRCh38
4647095 c.73C>T p.L25L Chr.2: 240891758-240891758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762926101 Chr.2: 240891881-240891881 on Build GRCh38
rs571841660 Chr.2: 240891783-240891783 on Build GRCh38
rs773413880 Chr.2: 240891785-240891785 on Build GRCh38
rs765424172 Chr.2: 240891875-240891875 on Build GRCh38
rs542666257 Chr.2: 240891951-240891951 on Build GRCh38
rs769392717 Chr.2: 240891855-240891855 on Build GRCh38
rs748804283 Chr.2: 240891940-240891940 on Build GRCh38
rs745313572 Chr.2: 240891849-240891849 on Build GRCh38
rs771010191 Chr.2: 240891871-240891871 on Build GRCh38
rs767419710 Chr.2: 240891999-240891999 on Build GRCh38
rs866673330 Chr.2: 240891864-240891864 on Build GRCh38
rs374993520 Chr.2: 240891794-240891794 on Build GRCh38
rs780682566 Chr.2: 240891913-240891913 on Build GRCh38
rs376900527 Chr.2: 240891985-240891985 on Build GRCh38
rs766628931 Chr.2: 240891788-240891788 on Build GRCh38
rs760831770 Chr.2: 240891786-240891786 on Build GRCh38
rs971983518 Chr.2: 240891968-240891968 on Build GRCh38
rs575994179 Chr.2: 240891997-240891997 on Build GRCh38
rs773501995 Chr.2: 240891867-240891867 on Build GRCh38
rs752718874 Chr.2: 240891811-240891811 on Build GRCh38
rs746812384 Chr.2: 240891840-240891840 on Build GRCh38
rs941739753 Chr.2: 240891919-240891919 on Build GRCh38
rs759504603 Chr.2: 240891874-240891874 on Build GRCh38
rs774719912 Chr.2: 240891856-240891856 on Build GRCh38
rs750160616 Chr.2: 240891917-240891917 on Build GRCh38
rs772586314 Chr.2: 240891865-240891865 on Build GRCh38
rs757195248 Chr.2: 240891831-240891832 on Build GRCh38
rs778274717 Chr.2: 240891961-240891961 on Build GRCh38
rs756900249 Chr.2: 240891843-240891843 on Build GRCh38
rs764131501 Chr.2: 240891884-240891884 on Build GRCh38
rs776963210 Chr.2: 240891872-240891872 on Build GRCh38
rs202007238 Chr.2: 240891782-240891782 on Build GRCh38
rs368268043 Chr.2: 240891988-240891988 on Build GRCh38
rs771368364 Chr.2: 240891971-240891971 on Build GRCh38
rs765064683 Chr.2: 240891805-240891805 on Build GRCh38
rs6709469 Chr.2: 240891879-240891879 on Build GRCh38
rs747354352 Chr.2: 240891965-240891965 on Build GRCh38
rs748714336 Chr.2: 240891861-240891861 on Build GRCh38
rs866990609 Chr.2: 240891908-240891908 on Build GRCh38
rs779563142 Chr.2: 240891927-240891927 on Build GRCh38
rs371391472 Chr.2: 240891793-240891793 on Build GRCh38
rs554554010 Chr.2: 240891966-240891966 on Build GRCh38
rs554284656 Chr.2: 240891873-240891873 on Build GRCh38
rs761060287 Chr.2: 240891870-240891870 on Build GRCh38
rs922874991 Chr.2: 240891878-240891878 on Build GRCh38
rs985761551 Chr.2: 240891882-240891882 on Build GRCh38
rs751354766 Chr.2: 240891897-240891897 on Build GRCh38
rs776712308 Chr.2: 240891983-240891983 on Build GRCh38
rs766386211 Chr.2: 240891836-240891836 on Build GRCh38
rs778971256 Chr.2: 240891904-240891905 on Build GRCh38
rs757094697 Chr.2: 240891904-240891904 on Build GRCh38
rs895703024 Chr.2: 240892000-240892000 on Build GRCh38
rs772529213 Chr.2: 240891957-240891957 on Build GRCh38
rs6737320 Chr.2: 240891847-240891847 on Build GRCh38
rs755568296 Chr.2: 240891921-240891921 on Build GRCh38

View Full Product Details