Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50915713-50915962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP15
Entrez Gene ID
9210
Gene Name
bone morphogenetic protein 15
Gene Aliases
GDF9B, ODG2, POF4
UniGene
Hs.532692
Gene Chromosome Location
Chr.X: 50910735-50916641 on Build GRCh38
Transcripts
NM_005448.2

SNP Detail

SNP ID Chromosome Location
rs781787652 Chr.X: 50915750-50915750 on Build GRCh38
rs782105156 Chr.X: 50915765-50915765 on Build GRCh38
rs782369768 Chr.X: 50915861-50915861 on Build GRCh38
rs782420643 Chr.X: 50915878-50915878 on Build GRCh38
rs782595246 Chr.X: 50915849-50915849 on Build GRCh38
rs782474336 Chr.X: 50915814-50915814 on Build GRCh38
rs1008418535 Chr.X: 50915828-50915828 on Build GRCh38
rs140427037 Chr.X: 50915773-50915773 on Build GRCh38
rs371418883 Chr.X: 50915841-50915841 on Build GRCh38
rs781824738 Chr.X: 50915741-50915741 on Build GRCh38
rs782483160 Chr.X: 50915853-50915853 on Build GRCh38
rs782687146 Chr.X: 50915839-50915839 on Build GRCh38
rs782799707 Chr.X: 50915771-50915771 on Build GRCh38
rs114823607 Chr.X: 50915871-50915871 on Build GRCh38
rs781858497 Chr.X: 50915822-50915822 on Build GRCh38
rs368260630 Chr.X: 50915858-50915858 on Build GRCh38
rs140319105 Chr.X: 50915862-50915862 on Build GRCh38
rs782459414 Chr.X: 50915934-50915934 on Build GRCh38
rs782281596 Chr.X: 50915881-50915881 on Build GRCh38
rs781874375 Chr.X: 50915902-50915902 on Build GRCh38
rs782780997 Chr.X: 50915938-50915938 on Build GRCh38
rs782705538 Chr.X: 50915795-50915795 on Build GRCh38
rs151186104 Chr.X: 50915837-50915837 on Build GRCh38
rs375806674 Chr.X: 50915844-50915844 on Build GRCh38
rs782621007 Chr.X: 50915821-50915821 on Build GRCh38
rs782731730 Chr.X: 50915928-50915928 on Build GRCh38
rs112760570 Chr.X: 50915809-50915809 on Build GRCh38
rs782069938 Chr.X: 50915908-50915908 on Build GRCh38
rs142156356 Chr.X: 50915780-50915780 on Build GRCh38
rs974301521 Chr.X: 50915883-50915883 on Build GRCh38

View Full Product Details