Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136261061-136261330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCLAF1
Entrez Gene ID
9774
Gene Name
BCL2 associated transcription factor 1
Gene Aliases
BTF, bK211L9.1
UniGene
Hs.486542
Gene Chromosome Location
Chr.6: 136256863-136290645 on Build GRCh38
Transcripts
NM_001077440.1 NM_001077441.1 NM_001301038.1 NM_014739.2 XM_005267237.3 XM_017011529.1 XM_017011530.1 XM_017011531.1 XM_017011532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3777135 c.2707G>A p.E903K Chr.6: 136261315-136261315 on Build GRCh38
5653193 c.2757+2T>A p.? Chr.6: 136261263-136261263 on Build GRCh38
3620929 c.2737G>A p.D913N Chr.6: 136261285-136261285 on Build GRCh38
3174904 c.2733delG p.K912fs*>9 Chr.6: 136261289-136261289 on Build GRCh38
594542 c.2701G>A p.E901K Chr.6: 136261321-136261321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111800140 Chr.6: 136261148-136261148 on Build GRCh38
rs745769824 Chr.6: 136261221-136261222 on Build GRCh38
rs537502804 Chr.6: 136261225-136261226 on Build GRCh38
rs376948280 Chr.6: 136261156-136261156 on Build GRCh38
rs946383743 Chr.6: 136261088-136261088 on Build GRCh38
rs777244187 Chr.6: 136261251-136261251 on Build GRCh38
rs777506614 Chr.6: 136261121-136261122 on Build GRCh38
rs747583650 Chr.6: 136261264-136261264 on Build GRCh38
rs752457268 Chr.6: 136261117-136261118 on Build GRCh38
rs777128127 Chr.6: 136261155-136261155 on Build GRCh38
rs755988618 Chr.6: 136261242-136261242 on Build GRCh38
rs562481221 Chr.6: 136261137-136261137 on Build GRCh38
rs779916409 Chr.6: 136261086-136261086 on Build GRCh38
rs193167212 Chr.6: 136261193-136261193 on Build GRCh38
rs570210520 Chr.6: 136261157-136261158 on Build GRCh38
rs751526906 Chr.6: 136261087-136261087 on Build GRCh38
rs769250966 Chr.6: 136261278-136261278 on Build GRCh38
rs769327751 Chr.6: 136261113-136261113 on Build GRCh38
rs761272577 Chr.6: 136261147-136261147 on Build GRCh38
rs770223828 Chr.6: 136261282-136261282 on Build GRCh38
rs780977812 Chr.6: 136261092-136261092 on Build GRCh38
rs769688973 Chr.6: 136261226-136261226 on Build GRCh38
rs754614460 Chr.6: 136261180-136261180 on Build GRCh38
rs763071484 Chr.6: 136261115-136261116 on Build GRCh38
rs745432277 Chr.6: 136261271-136261271 on Build GRCh38
rs754701506 Chr.6: 136261090-136261090 on Build GRCh38
rs768881257 Chr.6: 136261289-136261289 on Build GRCh38
rs766449517 Chr.6: 136261219-136261219 on Build GRCh38
rs766196853 Chr.6: 136261082-136261082 on Build GRCh38
rs1016876720 Chr.6: 136261205-136261205 on Build GRCh38
rs1002042606 Chr.6: 136261235-136261235 on Build GRCh38
rs375440569 Chr.6: 136261236-136261236 on Build GRCh38
rs774419180 Chr.6: 136261116-136261117 on Build GRCh38
rs370252607 Chr.6: 136261161-136261161 on Build GRCh38
rs759652247 Chr.6: 136261298-136261298 on Build GRCh38
rs76890371 Chr.6: 136261103-136261103 on Build GRCh38
rs774410839 Chr.6: 136261297-136261297 on Build GRCh38
rs766644301 Chr.6: 136261286-136261286 on Build GRCh38
rs62431282 Chr.6: 136261263-136261263 on Build GRCh38
rs748726912 Chr.6: 136261130-136261130 on Build GRCh38
rs763174649 Chr.6: 136261284-136261284 on Build GRCh38
rs778584534 Chr.6: 136261269-136261269 on Build GRCh38
rs749004030 Chr.6: 136261257-136261257 on Build GRCh38
rs770590143 Chr.6: 136261135-136261135 on Build GRCh38
rs112744301 Chr.6: 136261174-136261174 on Build GRCh38
rs62431283 Chr.6: 136261279-136261279 on Build GRCh38
rs756785936 Chr.6: 136261141-136261142 on Build GRCh38
rs777524158 Chr.6: 136261120-136261120 on Build GRCh38
rs752704429 Chr.6: 136261301-136261301 on Build GRCh38
rs781174370 Chr.6: 136261239-136261239 on Build GRCh38
rs773238687 Chr.6: 136261283-136261283 on Build GRCh38
rs758124399 Chr.6: 136261119-136261122 on Build GRCh38
rs751133979 Chr.6: 136261128-136261129 on Build GRCh38
rs1047141822 Chr.6: 136261158-136261158 on Build GRCh38
rs769442560 Chr.6: 136261112-136261113 on Build GRCh38
rs749046736 Chr.6: 136261286-136261288 on Build GRCh38
rs527483217 Chr.6: 136261221-136261221 on Build GRCh38
rs752403686 Chr.6: 136261241-136261241 on Build GRCh38
rs775830032 Chr.6: 136261113-136261114 on Build GRCh38
rs750678338 Chr.6: 136261157-136261157 on Build GRCh38

View Full Product Details