Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 129579969-129580222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP18
Entrez Gene ID
93663
Gene Name
Rho GTPase activating protein 18
Gene Aliases
MacGAP, SENEX, bA307O14.2
UniGene
Hs.486458
Gene Chromosome Location
Chr.6: 129576145-129710225 on Build GRCh38
Transcripts
NM_033515.2 XM_005267212.2 XM_005267213.1 XM_017011494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5802610 c.1865_1867delGAG p.G622delG Chr.6: 129580103-129580105 on Build GRCh38
5869272 c.1840G>A p.G614R Chr.6: 129580130-129580130 on Build GRCh38
5404338 c.1864G>A p.G622R Chr.6: 129580106-129580106 on Build GRCh38
6321211 c.1853C>A p.T618N Chr.6: 129580117-129580117 on Build GRCh38
3172694 c.1877delT p.L626fs*19 Chr.6: 129580093-129580093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752134236 Chr.6: 129580062-129580062 on Build GRCh38
rs752913693 Chr.6: 129580070-129580070 on Build GRCh38
rs201704062 Chr.6: 129580152-129580152 on Build GRCh38
rs138622213 Chr.6: 129580090-129580090 on Build GRCh38
rs111717033 Chr.6: 129580193-129580193 on Build GRCh38
rs866380047 Chr.6: 129580092-129580093 on Build GRCh38
rs202176182 Chr.6: 129580053-129580054 on Build GRCh38
rs767483798 Chr.6: 129580178-129580178 on Build GRCh38
rs777452965 Chr.6: 129580161-129580161 on Build GRCh38
rs746642535 Chr.6: 129580137-129580137 on Build GRCh38
rs763420217 Chr.6: 129580057-129580057 on Build GRCh38
rs756192472 Chr.6: 129580183-129580183 on Build GRCh38
rs763181823 Chr.6: 129580153-129580153 on Build GRCh38
rs749014790 Chr.6: 129580098-129580098 on Build GRCh38
rs774981728 Chr.6: 129580024-129580024 on Build GRCh38
rs781375429 Chr.6: 129580068-129580068 on Build GRCh38
rs754993303 Chr.6: 129580067-129580067 on Build GRCh38
rs373648411 Chr.6: 129580050-129580050 on Build GRCh38
rs900475372 Chr.6: 129580197-129580197 on Build GRCh38
rs770790096 Chr.6: 129580100-129580100 on Build GRCh38
rs368516363 Chr.6: 129580196-129580196 on Build GRCh38
rs756386744 Chr.6: 129580073-129580073 on Build GRCh38
rs893979296 Chr.6: 129580127-129580127 on Build GRCh38
rs372963329 Chr.6: 129580175-129580175 on Build GRCh38
rs945515028 Chr.6: 129580132-129580132 on Build GRCh38
rs372636892 Chr.6: 129580139-129580139 on Build GRCh38
rs188300275 Chr.6: 129580016-129580016 on Build GRCh38
rs771449468 Chr.6: 129580135-129580135 on Build GRCh38
rs760440321 Chr.6: 129580027-129580027 on Build GRCh38
rs766818758 Chr.6: 129580163-129580163 on Build GRCh38
rs116832521 Chr.6: 129580033-129580033 on Build GRCh38
rs377386270 Chr.6: 129580164-129580164 on Build GRCh38
rs760157465 Chr.6: 129580168-129580168 on Build GRCh38
rs768362090 Chr.6: 129580147-129580147 on Build GRCh38
rs775204763 Chr.6: 129580136-129580136 on Build GRCh38
rs1054110814 Chr.6: 129580115-129580115 on Build GRCh38
rs752715530 Chr.6: 129580179-129580179 on Build GRCh38
rs546947024 Chr.6: 129580004-129580004 on Build GRCh38
rs778665307 Chr.6: 129580101-129580101 on Build GRCh38
rs201029428 Chr.6: 129580059-129580059 on Build GRCh38
rs745866185 Chr.6: 129580124-129580124 on Build GRCh38
rs370714504 Chr.6: 129580167-129580167 on Build GRCh38
rs753558498 Chr.6: 129580103-129580105 on Build GRCh38
rs755722505 Chr.6: 129580054-129580054 on Build GRCh38

View Full Product Details