Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220067217-220067468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPNT1
Entrez Gene ID
10380
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
Gene Aliases
HEL20, PIP
UniGene
Hs.406134
Gene Chromosome Location
Chr.1: 220057482-220089916 on Build GRCh38
Transcripts
NM_001286149.1 NM_001286150.1 NM_001286151.1 NM_006085.5 XM_005272998.2 XM_005272999.4 XM_005273000.4 XM_005273004.2 XM_005273005.4 XM_006711114.3 XM_011509063.2 XM_011509064.2 XM_011509065.2 XM_017000043.1 XM_017000044.1 XM_017000045.1 XM_017000046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1295884 c.406C>G p.L136V Chr.1: 220067370-220067370 on Build GRCh38
904097 c.385C>A p.L129I Chr.1: 220067391-220067391 on Build GRCh38
3997255 c.452A>C p.N151T Chr.1: 220067324-220067324 on Build GRCh38
3966278 c.432A>G p.K144K Chr.1: 220067344-220067344 on Build GRCh38
5798885 c.445G>C p.V149L Chr.1: 220067331-220067331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985861638 Chr.1: 220067253-220067253 on Build GRCh38
rs773517122 Chr.1: 220067416-220067416 on Build GRCh38
rs377607501 Chr.1: 220067422-220067422 on Build GRCh38
rs908961578 Chr.1: 220067265-220067265 on Build GRCh38
rs758623038 Chr.1: 220067319-220067319 on Build GRCh38
rs745401669 Chr.1: 220067348-220067348 on Build GRCh38
rs369795710 Chr.1: 220067366-220067366 on Build GRCh38
rs75678851 Chr.1: 220067324-220067324 on Build GRCh38
rs765455507 Chr.1: 220067432-220067432 on Build GRCh38
rs1038545867 Chr.1: 220067395-220067395 on Build GRCh38
rs534520912 Chr.1: 220067429-220067429 on Build GRCh38
rs753007929 Chr.1: 220067305-220067305 on Build GRCh38
rs531821553 Chr.1: 220067279-220067279 on Build GRCh38
rs200184441 Chr.1: 220067256-220067256 on Build GRCh38
rs374500121 Chr.1: 220067404-220067404 on Build GRCh38
rs777485080 Chr.1: 220067320-220067320 on Build GRCh38
rs377435032 Chr.1: 220067270-220067270 on Build GRCh38
rs199772736 Chr.1: 220067392-220067392 on Build GRCh38
rs779308410 Chr.1: 220067301-220067301 on Build GRCh38
rs766906770 Chr.1: 220067284-220067284 on Build GRCh38
rs371343018 Chr.1: 220067430-220067430 on Build GRCh38
rs908755670 Chr.1: 220067252-220067252 on Build GRCh38
rs770775242 Chr.1: 220067331-220067331 on Build GRCh38
rs761034332 Chr.1: 220067421-220067421 on Build GRCh38
rs753972410 Chr.1: 220067295-220067295 on Build GRCh38
rs755051171 Chr.1: 220067300-220067300 on Build GRCh38
rs570830391 Chr.1: 220067438-220067438 on Build GRCh38
rs775078811 Chr.1: 220067255-220067255 on Build GRCh38
rs781045277 Chr.1: 220067333-220067333 on Build GRCh38
rs750780599 Chr.1: 220067274-220067274 on Build GRCh38
rs548349918 Chr.1: 220067383-220067383 on Build GRCh38

View Full Product Details