Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55287058-55287329 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRSK1
Entrez Gene ID
84446
Gene Name
BR serine/threonine kinase 1
Gene Aliases
hSAD1
UniGene
Hs.182081
Gene Chromosome Location
Chr.19: 55284101-55312562 on Build GRCh38
Transcripts
NM_032430.1 XM_005259327.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3371497 c.247G>A p.A83T Chr.19: 55287229-55287229 on Build GRCh38
2886821 c.300C>T p.Y100Y Chr.19: 55287282-55287282 on Build GRCh38
3423185 c.192C>T p.I64I Chr.19: 55287062-55287062 on Build GRCh38
3707412 c.258G>A p.K86K Chr.19: 55287240-55287240 on Build GRCh38
1001454 c.219G>A p.S73S Chr.19: 55287089-55287089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770212801 Chr.19: 55287127-55287127 on Build GRCh38
rs770577241 Chr.19: 55287142-55287142 on Build GRCh38
rs760341371 Chr.19: 55287089-55287089 on Build GRCh38
rs377180108 Chr.19: 55287203-55287203 on Build GRCh38
rs147404516 Chr.19: 55287164-55287164 on Build GRCh38
rs760266753 Chr.19: 55287138-55287138 on Build GRCh38
rs973482708 Chr.19: 55287167-55287167 on Build GRCh38
rs374368842 Chr.19: 55287166-55287166 on Build GRCh38
rs372144256 Chr.19: 55287246-55287246 on Build GRCh38
rs752672529 Chr.19: 55287163-55287163 on Build GRCh38
rs202159884 Chr.19: 55287207-55287207 on Build GRCh38
rs905688182 Chr.19: 55287256-55287256 on Build GRCh38
rs988415715 Chr.19: 55287120-55287120 on Build GRCh38
rs749804905 Chr.19: 55287131-55287131 on Build GRCh38
rs11878562 Chr.19: 55287197-55287197 on Build GRCh38
rs145026932 Chr.19: 55287080-55287080 on Build GRCh38
rs138223768 Chr.19: 55287187-55287187 on Build GRCh38
rs201641035 Chr.19: 55287252-55287252 on Build GRCh38
rs56384716 Chr.19: 55287209-55287209 on Build GRCh38
rs764723300 Chr.19: 55287149-55287149 on Build GRCh38
rs769343476 Chr.19: 55287208-55287208 on Build GRCh38
rs376192653 Chr.19: 55287134-55287134 on Build GRCh38
rs772696023 Chr.19: 55287128-55287129 on Build GRCh38
rs200202914 Chr.19: 55287145-55287145 on Build GRCh38
rs1044281093 Chr.19: 55287255-55287255 on Build GRCh38
rs773064177 Chr.19: 55287087-55287087 on Build GRCh38
rs182987265 Chr.19: 55287188-55287188 on Build GRCh38
rs537122207 Chr.19: 55287170-55287170 on Build GRCh38
rs777515166 Chr.19: 55287119-55287119 on Build GRCh38
rs753338237 Chr.19: 55287104-55287104 on Build GRCh38
rs201751771 Chr.19: 55287110-55287110 on Build GRCh38
rs544837132 Chr.19: 55287249-55287249 on Build GRCh38
rs748611163 Chr.19: 55287098-55287100 on Build GRCh38
rs369157869 Chr.19: 55287130-55287130 on Build GRCh38
rs753928719 Chr.19: 55287274-55287274 on Build GRCh38
rs781165135 Chr.19: 55287124-55287124 on Build GRCh38
rs745897153 Chr.19: 55287125-55287125 on Build GRCh38
rs202201193 Chr.19: 55287109-55287109 on Build GRCh38
rs757429135 Chr.19: 55287169-55287169 on Build GRCh38
rs778492767 Chr.19: 55287133-55287134 on Build GRCh38
rs1002330623 Chr.19: 55287257-55287257 on Build GRCh38
rs780759652 Chr.19: 55287129-55287129 on Build GRCh38
rs758256666 Chr.19: 55287114-55287114 on Build GRCh38
rs765855115 Chr.19: 55287092-55287092 on Build GRCh38
rs895699433 Chr.19: 55287229-55287229 on Build GRCh38
rs779450237 Chr.19: 55287194-55287194 on Build GRCh38
rs762912199 Chr.19: 55287221-55287221 on Build GRCh38
rs750593900 Chr.19: 55287185-55287185 on Build GRCh38
rs371764150 Chr.19: 55287192-55287192 on Build GRCh38
rs769125824 Chr.19: 55287083-55287083 on Build GRCh38
rs572232748 Chr.19: 55287235-55287235 on Build GRCh38
rs779418114 Chr.19: 55287282-55287282 on Build GRCh38
rs575462692 Chr.19: 55287276-55287276 on Build GRCh38
rs374646731 Chr.19: 55287195-55287195 on Build GRCh38
rs759143751 Chr.19: 55287146-55287146 on Build GRCh38
rs567004778 Chr.19: 55287121-55287121 on Build GRCh38
rs763956789 Chr.19: 55287228-55287228 on Build GRCh38
rs1021150227 Chr.19: 55287136-55287136 on Build GRCh38
rs571400619 Chr.19: 55287165-55287165 on Build GRCh38
rs776004859 Chr.19: 55287078-55287078 on Build GRCh38
rs376675554 Chr.19: 55287273-55287273 on Build GRCh38
rs141794317 Chr.19: 55287234-55287234 on Build GRCh38
rs199712894 Chr.19: 55287193-55287193 on Build GRCh38

View Full Product Details